Skip to Content
Merck
All Photos(2)

Key Documents

T6575

Sigma-Aldrich

TOFA

≥98% (HPLC)

Synonym(s):

5-(Tetradecyloxy)-2-furoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32O4
CAS Number:
Molecular Weight:
324.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

mp

112-115 °C

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCOc1ccc(o1)C(O)=O

InChI

1S/C19H32O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-16-22-18-15-14-17(23-18)19(20)21/h14-15H,2-13,16H2,1H3,(H,20,21)

InChI key

CZRCFAOMWRAFIC-UHFFFAOYSA-N

Application

TOFA has been used as a lipid biosynthesis inhibitor in mesenchymal stromal cells (MSCs), human pluripotent stem cells., an acetyl-CoA carboxylase 1 inhibitor in murine adipocyte cell lines., a lipolysis inhibitor in cancer stem cells (CSCs).
TOFA has been used as an ACC inhibitor to study its effect on insulin secretion in INS-1E cells.

Biochem/physiol Actions

5-(Tetradecyloxy)-2-furoic acid (TOFA) elicits hypolipidemic functionality by favoring fatty acid breakdown and at the same time preventing biosynthesis. It induces apoptosis in pancreatic cancer cells and favors tumor suppression.
Studies have reported that TOFA can inhibit the anorectic effect of subcutaneous tamoxifen (TMX) on food intake during refeeding experiments in rats.
TOFA is a cell-permeable, potent, reversible, and competitive inhibitor of acetyl-CoA carboxylase (ACC).
TOFA is a cell-permeable, potent, reversible, and competitive inhibitor of acetyl-CoA carboxylase (ACC). TOFA is a key enzyme involved in fatty acid biosynthesis. TOFA inhibits cellular fatty acid synthesis in a dose-dependent manner (IC50 = 4 μM in human breast cancer cell line MCF7).

Preparation Note

TOFA is soluble in DMSO at a concentration that is greater than 2 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetyl-CoA carboxylase 1-dependent protein acetylation controls breast cancer metastasis and recurrence
Garcia MR, et al.
Cell Metabolism, 26(6), 842-855 (2017)
Extracellular matrix mechanical cues regulate lipid metabolism through Lipin-1 and SREBP
Romani P, et al.
Nature Cell Biology, 21(3), 338-338 (2019)
P T Lange et al.
mBio, 9(4) (2018-07-19)
Gammaherpesviruses are oncogenic pathogens that persist in ~95% of the adult population. Cellular metabolic pathways have emerged as important regulators of many viral infections, including infections by gammaherpesviruses that require several lipid synthetic pathways for optimal replication. Liver X receptors
Inhibition of fatty acid synthesis induces apoptosis of human pancreatic cancer cells
Nishi K, et al.
Anticancer Research, 36(9), 4655-4660 (2016)
Weibo Zhou et al.
Cancer research, 63(21), 7330-7337 (2003-11-13)
C75, an inhibitor of fatty acid synthase (FAS), induces apoptosis in cultured human cancer cells. Its proposed mechanism of action linked high levels of malonyl-CoA after FAS inhibition to potential downstream effects including inhibition of carnitine palmitoyltransferase-1 (CPT-1) with resultant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service