Skip to Content
Merck
All Photos(1)

Key Documents

T3830

Sigma-Aldrich

Triciribine hydrate

≥97% (HPLC)

Synonym(s):

API-2, Akt/PKB Signaling Inhibitor-2, Inhibitor V, NSC 154020, TCN, VD-0002

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N6O4 · xH2O
CAS Number:
Molecular Weight:
320.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

tan

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.Cn1nc(N)c2cn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c4ncnc1c24

InChI

1S/C13H16N6O4.H2O/c1-18-11-7-5(10(14)17-18)2-19(12(7)16-4-15-11)13-9(22)8(21)6(3-20)23-13;/h2,4,6,8-9,13,20-22H,3H2,1H3,(H2,14,17);1H2/t6-,8-,9-,13-;/m1./s1

InChI key

SPGRLWCWHWRPHX-DOKXERMVSA-N

Gene Information

human ... AKT1(207)
mouse ... AKT1(11651)
rat ... AKT1(24185)

Application

Triciribine hydrate has been used to study the effect of diethyldithiocarbamate (DDC) on matrix metalloproteinase-1 (MMP-1) in hepatic stellate cells1. It has also been used to analyze ADAM 10 activation by (-)-epigallocatechin-3-gallate (EGCG) in N2a cells overexpressing Swedish mutant APP (SweAPP N2a cells)2.

Biochem/physiol Actions

Triciribine is a highly selective Akt/PKB inhibitor, that selectively inhibits the cellular phosphorylation/activation of Akt1/2/3.

Features and Benefits

This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Triciribine hydrate is soluble in DMSO at a concentration that is greater than 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camilla Evangelisti et al.
Journal of cellular physiology, 226(3), 822-831 (2010-09-22)
Over the past 20 years, survival rates of T-cell acute lymphoblastic leukemia (T-ALL) patients have improved, mainly because of advances in polychemotherapy protocols. Despite these improvements, we still need novel and less toxic treatment strategies targeting aberrantly activated signaling networks
Hiroaki Kamijo et al.
The Journal of investigative dermatology, 140(3), 636-644 (2019-08-30)
Whereas atopic dermatitis (AD) is considered as a T helper 2 (Th2)-centered disease, IL-17-producing Th (Th17) cells are also activated in AD lesional skin. However, the relationship between Th17 responses and Th2 responses in AD is still to be elucidated.
Rocío López de la Lama et al.
PloS one, 13(8), e0202971-e0202971 (2018-08-30)
Misconceptions, lack of knowledge, and negative attitudes towards sharks act as barriers preventing actions required to tackle threats to shark populations, limiting the success of global shark conservation initiatives. Peru, a major player for the international trade of shark products
Muhammad Miftahussurur et al.
PloS one, 11(12), e0166199-e0166199 (2016-12-03)
Information regarding Helicobacter pylori antibiotic resistance in Indonesia was previously inadequate. We assessed antibiotic susceptibility for H. pylori in Indonesia, and determined the association between virulence genes or genetic mutations and antibiotic resistance. We recruited 849 dyspeptic patients who underwent
A Griveau et al.
Oncogene, 35(38), 5033-5042 (2016-04-05)
Little is known about the biological role of the phospholipase A2 receptor (PLA2R1) transmembrane protein. In recent years, PLA2R1 has been shown to have an important role in regulating tumor-suppressive responses via JAK2 activation, but the underlying mechanisms are largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service