Skip to Content
Merck
All Photos(1)

Key Documents

T2815

Sigma-Aldrich

Transforming Growth Factor-β2 human

≥97% (electrophoresis), recombinant, expressed in NSO cells, powder, suitable for cell culture

Synonym(s):

hTGF-β2, TGF-β2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Transforming Growth Factor-β2 human, TGF-β2, recombinant, expressed in NSO cells, powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

≥97% (electrophoresis)

form

powder

potency

0.04 ng/mL ED50

quality

endotoxin tested

mol wt

25 kDa

packaging

pkg of 10 μg
pkg of 2 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

suitability

suitable for

UniProt accession no.

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The transforming growth factor-β2 (TGFB2) gene is mapped to human chromosome 1q41. The encoded protein is localized to extracellular matrix. TGF-β2 is a member of the TGF-β superfamily. TGF-β exists in three isoforms (TGF-β1, TGF-β2 and TGF-β3) and are known to share similar structure and function.

Application

Transforming Growth Factor-β2 human has been used in chondrogenic, adipogenic and osteogenic differentiation of mesenchymal stem cells. It has also been used to induce extracellular matrix accumulation in primary human trabecular meshwork cells and further to study the action of tetramethylpyrazine on the same.

Biochem/physiol Actions

TGF-β2 is known to stimulate cell growth, differentiation, migration and survival. TGF-β2 acts as a key regulator for immune homeostasis by controlling lymphocyte proliferation, embryogenesis, hematopoiesis and apoptosis. TGF-β is also associated with inflammation and wound healing and is considered as a strong growth inhibitor for keratinocyte cells. TGF-β is associated with the process of tumor development, progression and metastasis. TGF-β2 is a tumor suppressor in the early stages of carcinogenesis, but in the later stages acts as a tumor promoter by inducing epithelial-mesenchymal transition and stimulating angiogenesis. The TGF-β gene is overexpressed in glioma tumors and also in colon cancer, gastric cancer and cervical lesions.
Transforming growth factor-β2 (TGF-β2), like TGF-β1, is produced by many cell types and reported to be most concentrated in mammalian platelets.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.1 mg bovine serum albumin

Analysis Note

The biological activity is measured by its ability to inhibit the IL-4-dependent proliferation of mouse HT-2 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transforming growth factor β (TGF-β) and inflammation in cancer.
Bierie B and Moses HL
Cytokine & Growth Factor Reviews, 21.1, 49-59 (2010)
TGF-β1 and TGF-β2 abundance in liver diseases of mice and men
Dropmann A, et al.
Oncotarget, 7(15), 19499?19518-19499?19518 (2016)
The immunopharmacological properties of transforming growth factor β.
Le Y, et al.
International Immunopharmacology, 13-14, 1771-1782 (2005)
Loss-of-function mutations in TGFB2 cause a syndromic presentation of thoracic aortic aneurysm
Lindsay ME, et al.
Nature Genetics, 44(8), 922?927-922?927 (201)
Curcumin suppresses doxorubicin-induced epithelial?mesenchymal transition via the inhibition of TGF-β and PI3K/AKT signaling pathways in triple-negative breast cancer cells.
Chen WC, et al.
Journal of Agricultural and Food Chemistry, 61.48, 11817-11824 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service