Skip to Content
Merck
All Photos(1)

Key Documents

T2057

Sigma-Aldrich

Trequinsin hydrochloride

≥98%

Synonym(s):

HL 725

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27N3O3 · HCl
CAS Number:
Molecular Weight:
441.95
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

SMILES string

Cl[H].COc1cc2CCN3C(=O)N(C)C(\C=C3c2cc1OC)=N\c4c(C)cc(C)cc4C

InChI

1S/C24H27N3O3.ClH/c1-14-9-15(2)23(16(3)10-14)25-22-13-19-18-12-21(30-6)20(29-5)11-17(18)7-8-27(19)24(28)26(22)4;/h9-13H,7-8H2,1-6H3;1H/b25-22+;

InChI key

DTCZZBVPTHVXFA-OSMRDGEFSA-N

Gene Information

Application

Trequinsin has been used as a PDE3 inhibitor in rat juxtaglomerular cells. This study reported that trequinsin can enhance cellular cAMP content, forskolin-induced cAMP synthesis, and renin release in cells.

Biochem/physiol Actions

Trequinsin is a strong antihypertensive agent that has a hemodynamic profile similar to that of arteriolar dilators. Trequinsin can block platelet aggregation and also inhibit tissue factor expression in human endothelial cells,.
Phosphodiesterase III inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Rius et al.
Life sciences, 54(22), 1735-1743 (1994-01-01)
In primary cultured bovine adrenal chromaffin cells (BACC), pituitary adenylate cyclase activating polypeptide 1-38 (PACAP) produced a dose related increase in tyrosine hydroxylase (TH) Vmax when measured 48 hours after the beginning of the treatment; a significant increase was observed
R F Booth et al.
Biochemical pharmacology, 36(20), 3517-3521 (1987-10-15)
The discovery and structure-activity of a new class of renal artery phosphodiesterase inhibitors is reported, some of which are highly selective for the guanosine cyclic 3',5'-monophosphate phosphodiesterase. One of these compounds, 5,6-dihydro-8,9,11,12-tetramethoxy-1,3-dioxo-1H-benz[f]- isoquino [8,1,2- hij]quinazoline-2(3H)-carboxylic acid, ethyl ester (9), is
R Pillai et al.
The Journal of biological chemistry, 269(48), 30676-30681 (1994-12-02)
We have created a series of deletion mutants of a human cardiac cAMP phosphodiesterase in order to define sequences necessary for function and to identify residues required for inhibition by cGMP and by the drugs milrinone and trequinsin. These truncated
Jorge E Torres-López et al.
European journal of pharmacology, 519(1-2), 75-79 (2005-08-23)
The local peripheral (subcutaneous) injection of phosphodiesterase 3 inhibitor trequinsin dose-dependently enhanced formalin-evoked flinching during late second phase of this test. Treatment with the nitric oxide synthase inhibitor N-L-nitro-arginine methyl ester or guanylyl cyclase inhibitor 1-H-[1,2,4,]oxadiazolo[4,3-a]quinoxalin-1-one significantly reversed trequinsin-induced pronociceptive
M Kurjak et al.
The American journal of physiology, 276(6 Pt 1), G1521-G1530 (1999-06-11)
The effect of nitric oxide (NO) on the release of bombesin-like immunoreactivity (BLI) was examined in synaptosomes of rat small intestine. The NO donor S-nitroso-N-acetylpenicillamine (SNAP; 10(-7) to 10(-4) M) significantly stimulated BLI release. In the presence of the NO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service