Skip to Content
Merck
All Photos(2)

Key Documents

T2019

Sigma-Aldrich

Thebaine

powder

Synonym(s):

Paramorphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO3
CAS Number:
Molecular Weight:
311.37
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

solubility

alcohol: soluble 1 in 10 of alcohol
benzene: soluble 1 in 18 in benzene
chloroform: soluble 50 mg/ml, clear, yellow
diethyl ether: soluble 1 in 135 of ether
H2O: insoluble

SMILES string

[H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@]1(C5=CC=C2OC)c34

InChI

1S/C19H21NO3/c1-20-9-8-19-12-5-7-15(22-3)18(19)23-17-14(21-2)6-4-11(16(17)19)10-13(12)20/h4-7,13,18H,8-10H2,1-3H3/t13-,18+,19+/m1/s1

InChI key

FQXXSQDCDRQNQE-VMDGZTHMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thebaine has been used for the synthesis of neopinone/codeinone ring systems. Thebaine has also been used as a 1 mg/ml standard in methanol for urine thebaine analysis in individuals who had consumed poppy seeds.

Biochem/physiol Actions

The (-)-isomer of thebaine is the naturally-occurring constituent of opium that is a precursor of morphine. It has poor antinociceptive activity and induces convulsions. It binds to μ and δ opioid receptors in vitro.

Physical form

Alkaloid powder

Preparation Note

Thebaine dissolves in chloroform at 50 mg/ml to yield a clear, yellow solution. It soluble in alcohol, ether, and benzene at 1:10, 1: 135, and 1:18 parts respectively. However, thebaine does not dissolve in water.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Cassella et al.
Journal of analytical toxicology, 21(5), 376-383 (1997-09-01)
The consumption of poppy seeds in various foods may lead to a positive opiate result in urine subjected to testing for drugs of abuse. As a natural constituent of poppy seeds, thebaine was investigated as a possible marker for poppy
Morphine support studies. 2. Formation of the neopinone/codeinone ring system via intramolecular 1, 6-addition of an amino moiety to a dienyl ketone.
Toth, J. E., and Fuchs, P. L.
The Journal of Organic Chemistry, 51(13), 2594-2596 (1986)
Sooyeun Lee et al.
Forensic science international, 204(1-3), 115-118 (2010-06-19)
Opium poppy products are often illegally used for both recreational and medicinal purposes. In order to demonstrate the ingestion of opium poppy substances, morphine, codeine and their metabolites have been identified. However, morphine and codeine also originate from the ingestion
Jihyun Shim et al.
The journal of physical chemistry. B, 115(22), 7487-7496 (2011-05-14)
Despite being studied for over 30 years, a consensus structure-activity relationship (SAR) that encompasses the full range peptidic and nonpeptidic μ-opioid receptor ligands is still not available. To achieve a consensus SAR the Conformationally Sampled Pharmacophore (CSP) method was applied
Shahram Seidi et al.
Analytica chimica acta, 701(2), 181-188 (2011-08-02)
Opium determination is of great importance from toxicological and pharmaceutical standpoints. In present work, electromembrane extraction (EME) coupled with high-performance liquid chromatography (HPLC) and ultraviolet (UV) detection was developed for determination of thebaine as a natural alkaloid, in different matrices

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service