Skip to Content
Merck
All Photos(1)

Key Documents

T151

Sigma-Aldrich

(+)-Thalidomide

≥98% (HPLC), powder

Synonym(s):

R-(+)-2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O4
CAS Number:
Molecular Weight:
258.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

ligand

thalidomide

Assay

≥98% (HPLC)

form

powder

reaction suitability

reagent type: ligand

color

white to beige

solubility

DMSO: 15 mg/mL, clear

originator

Celgene

storage temp.

room temp

SMILES string

O=C1CC[C@@H](N2C(=O)c3ccccc3C2=O)C(=O)N1

InChI

1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)/t9-/m1/s1

InChI key

UEJJHQNACJXSKW-SECBINFHSA-N

Gene Information

Application

Thalidomide has been used to study its teratogenic effects in chicken embryos and human embryonic cells. This study reported that thalidomide causes limb defects by stabilizing PTEN, inhibiting the expression of Akt and activating caspase-dependent apoptosis. Thalidomide has also been used for studying glutathione mediated teratogenic resistance in mouse embryos.

Biochem/physiol Actions

(-)-Thalidomide selectively inhibits biosynthesis of tumor necrosis factor α (TNF-α). (R)-Thalidomide is called "safe enantiomer", but it can be converted in the body to (S)-isomer.

Features and Benefits

This compound was developed by Celgene. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Thalidomide is soluble in DMSO at a concentration that is greater than or equal to 20 mg/ml. It is insoluble in ethanol and water.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianhui Zhao et al.
Neurochemical research, 41(12), 3171-3180 (2016-08-31)
Opioid analgesics have less efficacy in diabetic neuropathy treatment, and tolerance often occurs after chronic usage. Given that thalidomide can potentiate the morphine efficacy in diabetic neuropathy treatment, we investigated the effects of intrathecal administrations of thalidomide on morphine tolerance
Jürgen Knobloch et al.
Molecular and cellular biology, 28(2), 529-538 (2008-01-08)
Thalidomide, a drug used for the treatment of multiple myeloma and inflammatory diseases, is also a teratogen that causes birth defects, such as limb truncations and microphthalmia, in humans. Thalidomide-induced limb truncations result from increased cell death during embryonic limb
Jürgen Knobloch et al.
Molecular pharmaceutics, 5(6), 1138-1144 (2009-05-13)
Thalidomide as an effective treatment for multiple myeloma and leprosy has also caused birth defects in thousands of children five decades ago particularly in Europe. Thus its use in humans remains limited. The rapid and fatal approval of thalidomide at
R J D'Amato et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(9), 4082-4085 (1994-04-26)
Thalidomide is a potent teratogen causing dysmelia (stunted limb growth) in humans. We have demonstrated that orally administered thalidomide is an inhibitor of angiogenesis induced by basic fibroblast growth factor in a rabbit cornea micropocket assay. Experiments including the analysis
Agata Kowalczyk et al.
Biosensors & bioelectronics, 167, 112446-112446 (2020-08-21)
Fast, simple in use and highly effective voltammetric enantiosensor dedicated for determination of thalidomide (TD) enantiomers (especially towards the toxic (S)-enantiomer) in blood plasma is still desirable. Here we have proven that newly synthesized chiral naphthalene diimide (NDI) derivatives are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service