Skip to Content
Merck
All Photos(1)

Key Documents

T1327

Sigma-Aldrich

Tissue Inhibitor of Metalloproteinase-3 human

recombinant, expressed in NSO cells, >95% (SDS-PAGE)

Synonym(s):

TIMP-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

apparent mol wt ~30 kDa

technique(s)

inhibition assay: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TIMP3(7078)

Looking for similar products? Visit Product Comparison Guide

General description

Tissue inhibitor of metalloproteinases 3 (TIMP3) is localized in the extracellular matrix (ECM) of epithelial cells associated with kidneys, eyes and lungs.[1] It is majorly secreted in retinal pigment epithelium (RPE).[2] TIMP3 gene is mapped to human chromosome 22q12.3[1] and is a CLOCK-dependent diurnal gene.[3] TIMP proteins display a three-lobed structure and have conserved cysteine residues.[2]

Biochem/physiol Actions

The TIMPs are endogenous inhibitors of the matrix metalloproteinases (MMPs). TIMP-3 inhibits ADAM-17 (TACE) at nanomolar concentrations and also inhibits other metalloproteinases (sheddases) that mediate the shedding of soluble receptors and other proteins from the surface of cells.
Tissue inhibitor of metalloproteinases 3 (TIMP3) is a matrix metalloproteinase inhibitor.[3] TIMP proteins comprise the N-terminal region, which aids in the matrix metalloproteinase interaction. The C-terminal region is crucial for extracellular matrix (ECM) interaction.[2] Elevated expression of TIMP3 favors apoptosis in cancer types.[3] Its interaction with the vascular endothelial growth factor (VEGFR-2) leads to the regulation of angiogenesis.[1] TIMP3 also aids in protection against ultra-violet (UV)-induced cellular responses.[3] Missense mutations in the TIMP3 gene are implicated in Sorsby′s fundus dystrophy (SFD).[1] Mutations in the TIMP3 gene also leads to increased accumulation of the protein resulting in the thickening of the Bruch membrane. This, in turn, reduces membrane permeability towards nutrients and metabolites.[2]

Physical form

Lyophilized from a 0.2 μm filtered solution in 25 mM Tris and 0.15 M sodium chloride, pH 7.5.

Analysis Note

The biological activity is measured by its ability to inhibit human MMP-2 hydrolysis of a peptide substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Ardeljan et al.
European journal of human genetics : EJHG, 21(10), 1152-1157 (2013-02-21)
Age-related macular degeneration (AMD) is a leading cause of irreversible central visual loss in the elderly. A recent genome-wide association studies (GWAS) reported that rs9621532 near the tissue inhibitor of metalloproteinase 3 (TIMP3)/synapsin III (SYN3) region of 22q12.3 is associated
Ruth Appeltant et al.
Animal science journal = Nihon chikusan Gakkaiho, 88(9), 1279-1290 (2017-01-27)
In vitro maturation (IVM) in serum causes hampered expansion of porcine cumulus-oocyte complexes (COCs) due to excessive alpha
Sunyoung Park et al.
International journal of molecular sciences, 20(4) (2019-02-20)
The human skin is the outermost physical barrier and has its own circadian machinery that works either cooperatively with the central clock, or autonomously. Circadian rhythms have been observed in many functions related to epidermal homeostasis including hydration and inflammation
Han-Li Huang et al.
Theranostics, 9(22), 6676-6689 (2019-10-08)
Tissue inhibitors of metalloproteinase 3 (TIMP3) are a major endogenous inhibitor of matrix metalloproteinase (MMPs) that inhibit tumor growth, invasion, metastasis and angiogenesis. In this study, we found that TIMP3 expression is associated with positive prognosis of colorectal cancer (CRC)
Sarah Naessens et al.
Human mutation, 40(5), 539-551 (2019-01-23)
Sorsby fundus dystrophy (SFD) is a macular degeneration caused by mutations in TIMP3, the majority of which introduce a novel cysteine. However, the exact molecular mechanisms underlying SFD remain unknown. We aimed to provide novel insights into the functional consequences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service