Skip to Content
Merck
All Photos(1)

Key Documents

T0953

Sigma-Aldrich

14-3-3 Tau/Theta histidine-tagged human

recombinant, expressed in E. coli, C-terminal, ≥90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3 tau, 14-3-3 theta, 14-3-3t, 1C5, Bap1, HS1, Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide, YWHAQ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~30 kDa (14-3-3 proteins)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... YWHAQ(10971)

Biochem/physiol Actions

14-3-3 proteins are involved in a multitude of biological processes and play a regulatory role in processes such as apoptotic cell death, mitogenic signal transduction, and cell cycle control. Co-immunoprecipitation experiments demonstrate the interaction of 14-3-3 τ/θ adaptor protein with various proteins (UCP3, PKCζ). 14-3-3 τ/θ is involved in cell cycle arrest and cellular differentiation. 14-3-3 τ/θ plays an active role in mediating Vpr-induced cell cycle arrest in human T cells.

Physical form

Supplied as a 0.2 μm filtered solution in 50 mM Tris-HCl, pH 7.6, 1 mM EDTA, 1 mM DTT, and 20% glycerol (w/v).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathematical biology in Biology Direct.
Andrei Yakovlev
Biology direct, 3, 1-1 (2008-01-18)
B Pierrat et al.
European journal of biochemistry, 267(9), 2680-2687 (2000-04-28)
Uncoupling proteins (UCPs) are members of the superfamily of the mitochondrial anion carrier proteins (MATP). Localized in the inner membrane of the organelle, they are postulated to be regulators of mitochondrial uncoupling. UCP2 and 3 may play an important role
14-3-3 isotypes facilitate coupling of protein kinase Cζ to Raf-1: negative regulation by 14-3-3 phosphorylation.
Van Der Hoeven, P.C., et al.
The Biochemical Journal, 15, 297-306 (2000)
José M A Moreira et al.
Molecular & cellular proteomics : MCP, 7(7), 1225-1240 (2008-04-02)
The 14-3-3 proteins constitute a family of highly conserved and broadly expressed multifunctional polypeptides that are involved in a variety of important cellular processes that include cell cycle progression, growth, differentiation, and apoptosis. Although the exact cellular function(s) of 14-3-3
14-3-3 proteins: active cofactors in cellular regulation by serine/threonine phosphorylation.
Guri Tzivion et al.
The Journal of biological chemistry, 277(5), 3061-3064 (2001-11-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service