Skip to Content
Merck
All Photos(2)

Key Documents

T0807

Sigma-Aldrich

3-(N,N-Dimethylmyristylammonio)propanesulfonate

≥99% (TLC), BioXtra

Synonym(s):

3-(N,N-Dimethyltetradecylammonio)propanesulfonate, 3-(Myristyldimethylammonio)propanesulfonate, N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, Myristyl sulfobetaine, SB3-14

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
363.60
Beilstein:
6419727
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

description

zwitterionic

Quality Level

product line

BioXtra

Assay

≥99% (TLC)

mol wt

micellar avg mol wt 30,200

aggregation number

83

technique(s)

electrophoresis: suitable
ion chromatography: suitable

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

CMC

0.1-0.4 mM (20-25°C)

solubility

H2O: 50 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

detection

SMILES string

CCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C19H41NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-17-20(2,3)18-16-19-24(21,22)23/h4-19H2,1-3H3

InChI key

BHATUINFZWUDIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(N,N-Dimethylmyristylammonio)propanesulfonate is a zwitterionic surfactant used in various electrostatic ion chromatography techniques. [1]
Zwitterionic detergent.

Application

3-(N,N-Dimethylmyristylammonio)propanesulfonate has been used in a study to assess a method for quantifying perchlorate in biological samples using CE and capacitively coupled contactless conductivity detection. [2] It has also been used in a study to investigate an approach for establishing adsorbents for elimination of water-soluble organic dyes. [3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Cook et al.
Analytical chemistry, 73(13), 3022-3027 (2001-07-27)
The retention mechanism of electrostatic ion chromatography (EIC) is currently under debate and is the focus of this paper. A comprehensive set of retention data has been obtained on a C18 column coated with the zwitterionic surfactant 3-(N,N-dimethylmyristylammonio)propanesulfonate used with
A novel adsorbent obtained by inserting carbon nanotubes into cavities of diatomite and applications for organic dye elimination from contaminated water
Yu, H. and B. Fugetsu
Journal of Hazardous Materials, 177, 8-8 (2010)
J T Poolman et al.
Antonie van Leeuwenhoek, 53(6), 413-419 (1987-01-01)
Outer membrane proteins (OMPs) were isolated from meningococcal strain H44/76 (B:15:P1.16) by detergent extraction of bacteria. A final product containing class 1 (P1.16), 3(15), 4 OMPs and 5% (w/w) lipooligosaccharide was obtained. Two experimental vaccines were prepared: OMP-detergent and OMP-C
G J Speijers et al.
Vaccine, 7(4), 364-368 (1989-08-01)
In order to assess the systemic toxicity of Zwittergent 3-14, a detergent used in the protein-detergent complex vaccines for its attractive immunogenic properties, a subacute toxicity study was performed. In this 4-week toxicity experiment five female and five male rats
Erick Sarmiento-Gomez et al.
The journal of physical chemistry. B, 114(38), 12193-12202 (2010-09-10)
We study the Brownian motion of probe particles embedded in a wormlike micellar fluid made of a zwitterionic surfactant N-tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate (TDPS), sodium dodecyl sulfate (SDS), and salty water to get structural and dynamical information of the micellar network. The motion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service