Skip to Content
Merck
All Photos(1)

Key Documents

SRP8049

Sigma-Aldrich

Periostin mouse

recombinant, expressed in CHO cells, FLAG® tagged, >95% (SDS-PAGE)

Synonym(s):

OSF-2, Osteoblast-specific factor 2, PC cell-derived growth factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in CHO cells

tag

FLAG® tagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~85 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Postn(50706)

General description

Periostin (POSTN) is also called as osteoblast-specific factor 2. It is a matricellular protein, located in the alveolar walls. This gene is located on human chromosome 13q13.
Periostin was originally isolated as a osteoblast-specific factor that functions as a cell adhesion molecule for preosteoblasts and is thought to be involved in osteoblast recruitment, attachment and spreading. Additionally, periostin expression has previously been shown to be significantly increased by both transforming growth factor beta1 (TGF-beta1) and bone morphogenetic protein (BMP-2). Abnormal expression of periostin is also linked to angiogenesis and metastasis in epithelial tumors.

Biochem/physiol Actions

Periostin (POSTN) participates in collagen fibrillogenesis. It plays a major role in the development of scar after traumatic spinal cord injury (SCI). Periostin controls the activities of fibrocyte to stimulate myofibroblast differentiation and lung fibrosis. It modulates the amount of collagen in tissue. Periostin also participates in the assembly of the extracellular matrix (ECM) architecture.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Mouse periostin (aa 24-783) (isoform 5) is fused at the C-terminus to a FLAG®-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai-Wen Yu et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 117(4), 292-300 (2018-01-08)
Appropriate mechanical stress plays an important role in regulating the proliferation and differentiation of osteoblasts, whereas high-level mechanical stress may be harmful and compromise cell survival. Periostin, a matricellular protein, is essential in maintaining functional integrity of bone and collagen-rich
Periostin deficiency causes severe and lethal lung injury in mice with bleomycin administration.
Kondoh H, et al.
The Journal of Histochemistry and Cytochemistry, 64(7), 441-453 (2016)
Shoichiro Ohta et al.
PloS one, 12(3), e0174547-e0174547 (2017-03-30)
The natural course of idiopathic pulmonary fibrosis (IPF) is variable. Predicting disease progression and survival in IPF is important for treatment. We previously demonstrated that serum periostin has the potential to be a prognostic biomarker for IPF. Our aim was
Nicolette M Jeanblanc et al.
Clinica chimica acta; international journal of clinical chemistry, 464, 228-235 (2016-10-19)
Periostin is being investigated as a potential biomarker for T-helper-2 (Th2)-driven asthma or eosinophilic inflammation and may help to identify patients more likely to benefit from interleukin-13-targeted treatments. We report the development and analytic performance of the investigational use only
Periostin regulates fibrocyte function to promote myofibroblast differentiation and lung fibrosis.
Ashley SL, et al.
Mucosal Immunology, 10(2), 341?351-341?351 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service