Skip to Content
Merck
All Photos(1)

Key Documents

SRP8026

Sigma-Aldrich

Annexin V human

recombinant, expressed in E. coli, GST tagged, ≥95% (SDS-PAGE)

Synonym(s):

Annexin A5, Calphobindin I, EC 2.7.1.23, Endonexin II, Placental anticoagulantprotein I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

tag

GST tagged

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

~52 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

activity assay: suitable

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ANXA5(308)

General description

Annexin V is a member of the annexin family of calcium and membrane-binding proteins that have anticoagulatory and anti-inflammatory properties. The encoded protein is 320 amino acid long and is folded into four domains, each having five α-helices, forming a superhelical globular structure. The ANX5 gene is mapped to human chromosome 4q26-q28 and spans a length of 28kb containing 13 exons and 12 introns.

Application

Annexin V has been used to examine phosphorylation activity of ChaK1 (channel-kinase 1).

Biochem/physiol Actions

Annexins are a family of water-soluble proteins that bind to membranes and phospholipids in a calcium-dependent manner. They participate in several cellular functions, such as membrane fusion and cytoskeletal interaction. Annexin V is a potent anticoagulant and serves as a voltage-gated calcium channel when bound to membrane in vitro. It is found to inhibit phospholipase A2 and protein kinase C.

Physical form

Lyophilized from PBS, pH 7.4.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Recombinant human annexin V fused to a GST-tag.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Huang et al.
Oxidative medicine and cellular longevity, 2020, 4921562-4921562 (2020-08-28)
Neuroinflammation plays an important pathological role in experimental surgical brain injury (SBI). Apoptotic associated with phosphatidylserine (PS) externalization promotes anti-inflammatory mediator TGF-β1 release. In the present study, we investigated the anti-neuroinflammation effect of PS liposome or isoflurane pretreatment via PS/CD36/TGF-β1
Shanyue Wang et al.
International immunopharmacology, 84, 106513-106513 (2020-04-25)
Ischemia-reperfusion (I/R) injury causes cardiac dysfunction through several mechanisms including oxidative stress and pro-inflammation. Eleutheroside E (EE) has protective effects in ischemia tissue and anti-inflammatory action. However, the effect of EE on I/R-injured cardiomyocytes is unknown. In this study, we
Zhiqiang Wei et al.
Chinese medicine, 15, 12-12 (2020-02-07)
Osteoarthritis (OA) is a metabolic disorder and able to be relieved by traditional Chinese medicines. However, the effect of Ligusticum wallichii on OA is unknown. Cytokine IL-1β and L. wallichii extracts were used to stimulate the primary mouse chondrocytes. MTT
Claire Pluchart et al.
Journal of thrombosis and thrombolysis, 48(2), 195-202 (2019-06-09)
Thromboembolic events are frequent and serious complications of acute lymphoblastic leukaemia treatment. The importance of chemotherapy in the pathogenesis of this increased risk is enhanced by the fact that thrombosis rarely occurs at diagnosis. Our study aims at investigating the
Shruti Sanjay Taparia et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 83, 130-140 (2016-10-25)
Over the last four centuries, cocoa and chocolate have been described as having potential medicinal value. As of today, Theobroma cacao L. (Sterculiaceae) and its products are consumed worldwide. They are of great research interest because of the concentration dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service