SRP5227
PAD6, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
PADI6
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~100 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... PADI6(353238)
General description
Peptidyl arginine deiminase 6 (PAD6) or PADI6 is a member of the peptidylarginine deiminases. It is expressed in oocytes and preimplantation embryos. The gene encoding this protein is localized on human chromosome 1p36.
Biochem/physiol Actions
Peptidyl arginine deiminases (PADs) convert arginine residues to citrulline residues in the presence of calcium ions. The PAD family members are thought to be involved in multiple sclerosis and rheumatoid arthritis pathophysiology, and they play a role in epidermis homeostasis. PAD6 is essential for formation of a novel oocyte-restricted fibrous structure, the cytoplasmic lattices (CPLs). PAD6/CPL superstructure plays a key role in regulating microtubule-mediated organelle positioning and movement.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Analysis Note
This protein is not assayed for enzymatic activity.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Potential Role of Peptidylarginine Deiminase Enzymes and Protein Citrullination in Cancer Pathogenesis
Biochemistry Research International (2012)
Regulation of mouse oocyte microtubule and organelle dynamics by PADI6 and the cytoplasmic lattices.
Developmental biology, 350(2), 311-322 (2010-12-15)
Organelle positioning and movement in oocytes is largely mediated by microtubules (MTs) and their associated motor proteins. While yet to be studied in germ cells, cargo trafficking in somatic cells is also facilitated by specific recognition of acetylated MTs by
BioEssays : news and reviews in molecular, cellular and developmental biology, 25(11), 1106-1118 (2003-10-28)
Peptidylarginine deiminase (PAD, EC 3.5.3.15) enzymes catalyze the conversion of protein-bound arginine to citrulline. This post-translational modification may have a big impact on the structure and function of the target protein. In this review, we will discuss the effects of
Acefylline activates filaggrin deimination by peptidylarginine deiminases in the upper epidermis.
Journal of Dermatological Science (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service