Skip to Content
Merck
All Photos(1)

Key Documents

SRP3327

Sigma-Aldrich

IL-1 RECEPTOR ANTAGONIST human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

ICIL-1RA, IL-1RN, IL-1Ra, IRAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20-40 ng/mL

mol wt

17.2 kDa

packaging

pkg of 100 μg

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

technique(s)

activity assay: suitable

impurities

endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1R1(3554)

General description

Research area: Immunology & cytokines

Interleukin-1 receptor antagonist (IL-1RA) is a naturally occurring inflammatory inhibitor protein. IL-1Ra is produced by corneal epithelial cells, monocytes, neutrophils, macrophages, and fibroblasts. Recombinant human IL-1Ra is a 17.2kDa protein consisting of 153 amino acid residues.

Application

Interleukin-1 receptor antagonist (IL-1RA) human has been used:
  • to study the role of interleukin (IL)-1β receptor in cell senescence induced by doxorubicin
  • to block the IL-1β binding to its receptor
  • to understand the role of autocrine interleukin-1β (IL-1β) signaling downstream of cl- in RPS27 modulation

Biochem/physiol Actions

Interleukin-1 receptor antagonist (IL-1RA) inhibits the activity of IL-1α and IL-1β by competitively blocking binding to their type I and type II receptors. Therapeutically, IL-1RA may help in the treatment of sepsis, cachexia, rheumatoid arthritis, chronic myelogenous leukemia, asthma, psoriasis, and inflammatory bowel disease.

Physical form

Lyophilized from 20 mM Tris, pH 8.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The role of cytokines in cancer-related fatigue.
Kurzrock R
Cancer, 92, 1684-1688 (2001)
Exposure of T lymphocytes to leflunomide but not to dexamethasone favors the production by monocytic cells of interleukin-1 receptor antagonist and the tissue-inhibitor of metalloproteinases-1 over that of interleukin-1beta and metalloproteinases.
Deage V
European Cytokine Network, 9, 663-668 (1998)
Association of IL-1a rs17561 and IL-1 RN rs315952 polymorphisms with Tourette syndrome: a family-based study.
He F
International Journal of Clinical and Experimental Pathology, 8, 4182-4185 (2015)
Jenaro A Espitia-Corredor et al.
Biochimica et biophysica acta. Molecular basis of disease, 1868(11), 166525-166525 (2022-08-21)
Cardiac fibroblasts (CFs) undergo senescence in reaction to different stressors, leading to a poor prognosis of cardiac disease. Doxorubicin (Doxo) is an antineoplastic drug with strong cardiotoxic effects, which induces IL-1β secretion and thus, triggers a potent pro-inflammatory response. Doxo
Relationship of plasma leptin to plasma cytokines and human survivalin sepsis and septic shock.
Arnalich F
The Journal of Infectious Diseases, 180, 908-911 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service