SRP2005
TFIIE β, p34 human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Synonym(s):
FE, TF2E2, TFIIE-B
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.26
biological source
human
recombinant
expressed in E. coli
Assay
≥70% (SDS-PAGE)
form
frozen liquid
mol wt
~35 kDa
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
concentration
250 μg/mL
technique(s)
western blot: suitable
color
clear colorless
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... GTF2E2(2961)
Biochem/physiol Actions
The human Transcription Factor IIE (TFIIE) is composed of 56 kDa ( α) and 34 kDa ( β) subunits and is shown to be a heterotetramer. The 56-kDa subunit contains a region similar to a zinc-binding domain and a region sharing homology with the catalytic loop of a kinase domain. TFIIE binds to RNA polymerase II in solution and joins the pre-initiation complex probably concomitant with RNA polymerase II and TFIIF.
Physical form
Clear and colorless frozen liquid solution
Preparation Note
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Inostroza et al.
The Journal of biological chemistry, 266(14), 9304-9308 (1991-05-15)
Mammalian RNA polymerase II transcription factor IIE (TFIIE) was purified to apparent homogeneity. The activity copurified with polypeptides of 34 and 56 kDa. The 56-kDa subunit was sufficient for low levels of transcription activity in a transcription system reconstituted in
Y Ohkuma et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(23), 9163-9167 (1990-12-01)
Human transcription factor TFIIE, a ubiquitous factor required for transcription initiation by RNA polymerase II, was purified to homogeneity by a combination of conventional and HPLC steps. The purified TFIIE contained equimolar amounts of 57-kDa (TFIIE-alpha) and 34-kDa (TFIIE-beta) polypeptides
M G Peterson et al.
Nature, 354(6352), 369-373 (1991-12-05)
The general transcription factor IIE (TFIIE) is an essential component of the eukaryotic RNA polymerase II initiation complex. We have isolated human complementary DNA clones for both the subunits of TFIIE. Using purified recombinant proteins we find that both subunits
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service