Skip to Content
Merck
All Photos(1)

Key Documents

SRP0203

Sigma-Aldrich

Peroxiredoxin II Active human

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

Synonym(s):

PRDX2, PRP, Thioredoxin-dependent peroxide reductase 1, NKEF-B, Natural killer cell-enhancing factor B, TDPX1, Thiol-specific antioxidant 1, Thioredoxin peroxidase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

specific activity

≥149 pmol/min-μg

mol wt

22.7 kDa

packaging

pkg of 100 μg

concentration

>0.02 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRDX2(7001)

General description

Research area: CELL SIGNALING

Peroxiredoxin 2 (PRDX2) is a typical peroxiredoxin, with 2-cysteine (Cys), a catalytic Cys51 and a resolving cysteine Cys172. Peroxiredoxins are peroxides reducing enzymes, and 2-Cys Prx subfamily contains four members in mammals, including PRDX1-4. PRDX2 is the predominant peroxidase present in mammalian erythrocytes, and its nitrated form is present in brains of patients with early Alzheimer’s disease.

Application

Peroxiredoxin II Active human has been used as a supplement in the IVM (in vitro maturation) medium for the IVM of bovine follicular oocytes.

Biochem/physiol Actions

Peroxiredoxin-2 is a ubiquitous redox-active intracellular enzyme that acts as a redox-dependent inflammatory mediator, triggering macrophages to produce and release TNF-α (tumor necrosis factor). It may be involved in peroxidase activity and resistance to overoxidation. Its levels are elevated in several human cancer cells and tissues, including colorectal cancer (CRC) and it influences diverse cellular processes involving cells survival, proliferation and apoptosis. Prdx2 plays an essential role in regulating oxidation-induced apoptosis in CRC cells and may have potential as a therapeutic target in CRC. It may also act as a possible candidate biomarker for pancreatic cancer. Prx2 is a key regulator of invasion and metastasis in melanoma.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of peroxiredoxin II on the quality and mitochondrial activity of pre-implantation bovine embryos.
Fakruzzaman M et al
Animal Reproduction Science, 159, 172-183 (2015)
Lía M Randall et al.
The Journal of biological chemistry, 289(22), 15536-15543 (2014-04-11)
Peroxiredoxins (Prx) are efficient thiol-dependent peroxidases and key players in the mechanism of H2O2-induced redox signaling. Any structural change that could affect their redox state, oligomeric structure, and/or interaction with other proteins could have a significant impact on the cascade
Linkage of inflammation and oxidative stress via release of glutathionylated peroxiredoxin-2, which acts as a danger signal.
Salzano S, Checconi P, Hanschmann EM, et al.
Proceedings of the National Academy of Sciences of the USA, 111(33), 12157-12162 (2014)
Weidong Lu et al.
Molecular and cellular biochemistry, 387(1-2), 261-270 (2013-11-16)
Peroxiredoxin 2 (Prdx2) is a member of the peroxiredoxin family, which is responsible for neutralizing reactive oxygen species. Prdx2 has been found to be elevated in several human cancer cells and tissues, including colorectal cancer (CRC), and it influences diverse
Doo Jae Lee et al.
Cancer research, 73(15), 4744-4757 (2013-06-12)
In melanoma, transition to the vertical growth phase is the critical step in conversion to a deadly malignant disease. Here, we offer the first evidence that an antioxidant enzyme has a key role in this transition. We found that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service