Skip to Content
Merck
All Photos(1)

Key Documents

SML3682

Sigma-Aldrich

ST2825

≥98% (HPLC)

Synonym(s):

(2R,4′R,8′aR)-1-[2-[4-[[2-(2,4-Dichlorophenoxy)acetyl]amino]phenyl]acetyl]tetrahydro-6′-oxo-spiro[pyrrolidine-2,7′(6′H)-[2H]pyrrolo[2,1-b][1,3]thiazine]-4′-carboxamide, PAM4-SP19-Beta8-NH2, ST 2825, ST-2825

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28Cl2N4O5S
CAS Number:
Molecular Weight:
591.51
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active peptidomimetic MyD88 homodimerization inhibitor with in vitro and in vivo efficacy against IL-1R and TLR9-dependent processes.
ST2825 is an orally active peptidomimetic of a heptapeptide structure in the MyD88 TIR BB-loop. ST2825 prevents IL-1beta-mediated activation of NF-kappaB by blocking MyD88 homodimerization and thereby the recruitment of IL-1 receptor-associated kinases IRAK1/4. ST2825 at 10 µg/mL completely suppresses 2.5 µg/mL TLR9 ligand ODN 2006-induced B-cell plasma cell differentation and antibodies production from PBMCs. ST2825 inhibits IL-1β-induced IL-6 production in mice (100 & 200 mg/kg p.o. prior to 20 µg/kg IL-1β i.p.) and protects against left ventricular (LV) dilatation and hypertrophy (25 mg/kg/day i.p.) in a murine model of nonreperfused acute myocardial infarction (AMI) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induction of cyclooxygenase-2 gene by Candida albicans through EGFR, ERK, and p38 pathways in human urinary epitheli
Medical Mycology, 55(3), 314-322 (2017)
Inhibition of myeloid differentiation factor 88(MyD88) by ST2825 provides neuroprotection after experimental traumatic brain injury in mice
Brain Research, 1643, 130-139 (2016)
Effect of ST2825 on the proliferation and apoptosis of human hepatocellular carcinoma cells
Genetics and molecular research : GMR, 15(1), 15016826-15016826 (2016)
Pharmacological inhibition of TLR9 activation blocks autoantibody production in human B cells from SLE patients
Rheumatology (Oxford, England), 49(12), 2281-2289 (2010)
Short-term MyD88 inhibition ameliorates cardiac graft rejection and promotes donor-specific hyporesponsiveness of skin grafts in mice
Transplant International : Official Journal of the European Society For Organ Transplantation, 29(8), 941-952 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service