Skip to Content
Merck
All Photos(1)

Key Documents

SML3637

Sigma-Aldrich

YM-254890

≥95% (HPLC)

Synonym(s):

N-Acetyl-L-threonyl-(aR)-a-hydroxybenzenepropanoyl-2,3-didehydro-N-methylalanyl-L-alanyl-N-methyl-L-alanyl-(3R)-3-[[(2S,3R)-2-(acetylamino)-3-hydroxy-4-methyl-1-oxopentyl]oxy]-L-leucyl-N,O-dimethyl-L-threonine (7→1)-lactone, QS 3666A, YM 254890

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H69N7O15
CAS Number:
Molecular Weight:
960.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

YM-254890, a cyclic depsipeptide isolated from the culture broth of Chromobacterium sp., is a cell-permeable, potent and selective inhibitor of Gαq/11 protein. YM-254890 specifically inhibits the GDP/GTP exchange by blocking GDP release from the Gq protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Kuschak et al.
British journal of pharmacology, 177(8), 1898-1916 (2019-12-28)
G proteins are intracellular switches that transduce and amplify extracellular signals from GPCRs. The Gq protein subtypes, which are coupled to PLC activation, can act as oncogenes, and their expression was reported to be up-regulated in cancer and inflammatory diseases.
Akiyuki Nishimura et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(31), 13666-13671 (2010-07-20)
Heterotrimeric GTP-binding proteins (G proteins) transmit extracellular stimuli perceived by G protein-coupled receptors (GPCRs) to intracellular signaling cascades. Hundreds of GPCRs exist in humans and are the targets of a large percentage of the pharmaceutical drugs used today. Because G
Jun Takasaki et al.
The Journal of biological chemistry, 279(46), 47438-47445 (2004-09-02)
YM-254890, which was isolated from the culture broth of Chromobacterium sp., inhibits ADP-induced platelet aggregation and has antithrombotic and thrombolytic effects. YM-254890 blocks Galpha(q/11)-coupled ADP receptor P2Y1-mediated Ca(2+) mobilization. Here we report that YM-254890 is a selective Galpha(q/11) inhibitor. YM-254890

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service