Skip to Content
Merck
All Photos(1)

Key Documents

SML3328

Sigma-Aldrich

Methyl protodioscin

≥98% (HPLC)

Synonym(s):

(3β,​22α,​25R)​-​26-​(β-​D-​Glucopyranosyloxy)​-​22-​methoxyfurost-​5-​en-​3-​yl O-​6-​deoxy-​α-​L-​mannopyranosyl-​(1→2)​-​O-​[6-​deoxy-​α-​L-​mannopyranosyl-​(1→4)​]​-β-​D-​glucopyranoside, 22-O-Methylprotodioscin, Methylprotodioscin, NSC 698790

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H86O22
CAS Number:
Molecular Weight:
1063.23
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

InChI

1S/C52H86O22/c1-21(20-66-46-40(61)39(60)36(57)31(18-53)70-46)10-15-52(65-7)22(2)33-30(74-52)17-29-27-9-8-25-16-26(11-13-50(25,5)28(27)12-14-51(29,33)6)69-49-45(73-48-42(63)38(59)35(56)24(4)68-48)43(64)44(32(19-54)71-49)72-47-41(62)37(58)34(55)23(3)67-47/h8,21-24,26-49,53-64H,9-20H2,1-7H3/t21-,22+,23+,24+,26+,27-,28+,29+,30+,31-,32-,33+,34+,35+,36-,37-,38-,39+,40-,41-,42-,43+,44-,45-,46-,47+,48+,49-,50+,51+,52-/m1/s1

InChI key

HSSJYSJXBOCKQM-GVTGEURHSA-N

Biochem/physiol Actions

Methyl protodioscin is a saponin isolated from medical plant Dioscoreaceae that exhibit anticancer properties. It appears that Methyl protodioscin inhibits proliferation and promoted apoptosis of pancreatic cancer through suppression of glycolysis. Methyl protodioscin protects rat brain from I/R injury through down-regulation Mul1 (Mitochondrial E3 ubiquitin ligase 1) and elevation of SOD2 (superoxide dismutase 2) protein levels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service