Skip to Content
Merck
All Photos(1)

Key Documents

SML3325

Sigma-Aldrich

Sob-AM2

≥98% (HPLC)

Synonym(s):

2-[4-[[4-Hydroxy-3-(1-methylethyl)phenyl]methyl]-3,5-dimethylphenoxy]-N-methylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO3
CAS Number:
Molecular Weight:
341.44
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C1=C(C=CC(CC2=C(C=C(C=C2C)OCC(NC)=O)C)=C1)O)C

InChI

1S/C21H27NO3/c1-13(2)18-10-16(6-7-20(18)23)11-19-14(3)8-17(9-15(19)4)25-12-21(24)22-5/h6-10,13,23H,11-12H2,1-5H3,(H,22,24)

InChI key

UWVFEENBJKSTRD-UHFFFAOYSA-N

Related Categories

General description

Sob-AM2 is a methyl amide derivative and a central nervous system (CNS)- selective prodrug of Sobetirome. Even in the absence of monocarboxylate transporter 8 (Mct8), Sob-AM2 can penetrate the brain and exhibit thyromimetic effects. It can promote extensive myelin repair in the brain and spinal cord of mouse models with demyelination. Sob-AM2 and other thyromimetics may also stimulate triggering receptor expressed on myeloid cell-2 (TREM2) expression in macrophages and microglia.

Biochem/physiol Actions

CNS-penetrating thyromimetic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective Thyroid Hormone Receptor-Beta (TR?) Agonists: New Perspectives for the Treatment of Metabolic and Neurodegenerative Disorders
Saponaro F, et al.
Frontiers in medicine (2020)
Soledad Bárez-López et al.
Thyroid : official journal of the American Thyroid Association, 28(9), 1211-1220 (2018-05-31)
Loss of function mutations in the thyroid hormone (TH)-specific cell membrane transporter, the monocarboxylate transporter 8 (MCT8), lead to profound psychomotor retardation and abnormal TH serum levels, with low thyroxine (T4) and high triiodothyronine (T3). Several studies point to impaired
Skylar J Ferrara et al.
Cell chemical biology, 29(2), 239-248 (2021-08-11)
Triggering receptor expressed on myeloid cells-2 (TREM2) is a cell surface receptor on macrophages and microglia that senses and responds to disease-associated signals to regulate the phenotype of these innate immune cells. The TREM2 signaling pathway has been implicated in
P Chaudhary et al.
Journal of neuroimmunology, 352, 577468-577468 (2021-01-11)
We have previously demonstrated that thyromimetics stimulate oligodendrocyte precursor cell differentiation and promote remyelination in murine demyelination models. We investigated whether a thyroid receptor-beta selective thyromimetic, sobetirome (Sob), and its CNS-targeted prodrug, Sob-AM2, could prevent myelin and axonal degeneration in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service