SML3259
Morroniside
≥95% (HPLC)
Synonym(s):
[1S-(1α,3β,4aβ,8β,8aβ)]-8-(β-D-Glucopyranosyloxy)-4,4a,8,8a-tetrahydro-3-hydroxy-1-methyl-1H,3H-Pyrano[3,4-c]pyran-5-carboxylic Acid Methyl Ester, b-Morroniside
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Biochem/physiol Actions
Morroniside is a bioactive natural product with antioxidant and anti-inflammatory activity. Its activities have included protecttion against cerebral ischemia/reperfusion injury by inhibiting neuron apoptosis and MMP2/9 expression and regulation of hair growth and cycle transition via activation of the Wnt/β-catenin signaling pathway.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Scientific reports, 8(1), 13785-13785 (2018-09-15)
Hair loss is characterized by a shortened hair anagen phase and hair follicles (HF) miniaturization. Morroniside is the most abundant iridoid glycoside extracted from Cornus officinalis and has various bioactivities in different cell functions and tissue regeneration. In this study
European journal of pharmacology, 834, 295-304 (2018-07-31)
Amyloid-β peptide (Aβ) plays a causal role in the development and progression of Alzheimer's disease (AD). Oxidative stress and activation of mitogen-activated protein kinase (MAPK) are involved in Aβ-induced neurotoxicity. Morroniside, one active monomer of dry ripe sarcocarp of Cornus
Experimental and therapeutic medicine, 16(3), 2229-2234 (2018-09-07)
The aim of the present study was to investigate the effect of morroniside against matrix metalloproteinase (MMP)2/9 and focal cerebral ischemia/reperfusion (I/R) injury in rats. A rat model of focal cerebral I/R injury rats was established and rats were administered
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service