Skip to Content
Merck
All Photos(1)

Key Documents

SML2786

Sigma-Aldrich

WRG-28

≥98% (HPLC)

Synonym(s):

DDR2 inhibitor WRG-28, N-Ethyl-4-[[(3-oxo-3H-phenoxazin-7-yl)oxy]methyl]-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N2O5S
CAS Number:
Molecular Weight:
410.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to very dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

WRG-28 is a potent, selective and extracellularly acting allosteric inhibitor of discoidin domain receptor 2 (DDR2) that potently inhibits invasion and migration in mice model of breast cancer. WRG-28 inhibits metastatic breast tumor cell colonization in the lungs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saumya S Gurbani et al.
Tomography (Ann Arbor, Mich.), 5(1), 53-60 (2019-03-12)
Histone deacetylases regulate a wide variety of cellular functions and have been implicated in redifferentiation of various tumors. Histone deacetylase inhibitors (HDACi) are potential pharmacologic agents to improve outcomes for patients with gliomas. We assessed the therapeutic efficacy of belinostat
Pengwei Lu et al.
Artificial cells, nanomedicine, and biotechnology, 47(1), 3955-3960 (2019-10-02)
Belinostat is a histone deacetylase inhibitor drug capable of regulating cell growth in diverse cancers. Nonetheless, little information clarified the role of Belinostat in breast cancer. Hence, the functions of Belinostat in breast cancer cells survival was disclosed in this
Reece G Kenny et al.
Journal of inorganic biochemistry, 206, 110981-110981 (2020-02-24)
Vorinostat (suberoylanilide hydroxamic acid; SAHA) and Belinostat are two hydroxamate-based histone deacetylase inhibitors that are used clinically as potent anti-cancer agents. Their metabolic breakdown into inactive metabolites such as carboxylic acid and glucuronic derivatives results in them having short half-lives
Li Ren Kong et al.
Nature communications, 11(1), 2086-2086 (2020-05-01)
Gain of function (GOF) DNA binding domain (DBD) mutations of TP53 upregulate chromatin regulatory genes that promote genome-wide histone methylation and acetylation. Here, we therapeutically exploit the oncogenic GOF mechanisms of p53 codon 158 (Arg158) mutation, a DBD mutant found
Xiaoyan Qiu et al.
Cellular reprogramming, 22(1), 14-21 (2020-02-06)
To improve the isolation efficiency of parthenogenetic embryonic stem cells (pESCs) in mice, it is necessary to optimize the method to increase in vitro developmental competence of mice parthenogenetic blastocysts. Therefore, this study aims to investigate an optimal method for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service