Skip to Content
Merck
All Photos(1)

Key Documents

SML2594

Sigma-Aldrich

Sapacitabine

Synonym(s):

CS 682, CYC 682, N-[1-(2-Cyano-2-deoxy-β-D-arabinofuranosyl)-1,2-dihydro-2-oxo-4-pyrimidinyl]hexadecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H42N4O5
CAS Number:
Molecular Weight:
490.64
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N1\C(=N\C(=O)CCCCCCCCCCCCCCC)\C=CN(C1=O)[C@@H]2O[C@@H]([C@H]([C@@H]2C#N)O)CO

InChI

1S/C26H42N4O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-23(32)28-22-16-17-30(26(34)29-22)25-20(18-27)24(33)21(19-31)35-25/h16-17,20-21,24-25,31,33H,2-15,19H2,1H3,(H,28,29,32,34)/t20-,21+,24-,25+/m0/s1

InChI key

LBGFKUUHOPIEMA-PEARBKPGSA-N

Biochem/physiol Actions

Sapacitabine is an orally active prodrug of nucleoside analog CNDAC that exhibits potent anti-tumor activity in both blood and solid tumors. CNDAC incorporates into DNA where it causes single-strand breaks (SSBs), which are converted into double strand breaks (DSBs) during S phase.
orally active prodrug of nucleoside analog CNDAC that incorporates into DNA where it causes single-strand breaks

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service