Skip to Content
Merck
All Photos(1)

Key Documents

SML2319

Sigma-Aldrich

TAT-Gap19 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

L-Tyrosylglycyl-L-arginyl-L-lysyl-L-lysyl-L-arginyl-L-arginyl-L-glutaminyl-L-arginyl-L-arginyl-L-arginyl-L-lysyl-L-glutaminyl-L-isoleucyl-L-?-glutamyl-L-isoleucyl-L-lysyl-L-lysyl-L-phenylalanyl-L-lysine trifluoroacetate salt, YGRKKRRQRRRKQIEIKKFK trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C119H212N46O26 · xC2HF3O2
CAS Number:
Molecular Weight:
2703.25 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

form

lyophilized powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

TAT-Gap19 is a cell penetrant analog of a connexin 43 (Cx43) hemichannels selective blocker Gap19. TAT-Gap19 inhibits TAT-Gap19 in mice brain without affecting gap junction channels.
cell penetrant analog of a connexin 43 (Cx43) hemichannels selective blocker Gap19

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Bazzigaluppi et al.
Neurobiology of disease, 101, 1-7 (2016-12-23)
Extracellular potassium concentration, [K+]o, is a major determinant of neuronal excitability. In the healthy brain, [K+]o levels are tightly controlled. During seizures, [K+]o increases up to 15mM and is thought to cause seizures due to its depolarizing effect. Although astrocytes
Verónica Abudara et al.
Frontiers in cellular neuroscience, 8, 306-306 (2014-11-07)
In the brain, astrocytes represent the cellular population that expresses the highest amount of connexins (Cxs). This family of membrane proteins is the molecular constituent of gap junction channels and hemichannels that provide pathways for direct cytoplasm-to-cytoplasm and inside-out exchange
Laura Walrave et al.
Frontiers in cellular neuroscience, 10, 288-288 (2017-01-10)
Astrocytes are active players in higher brain function as they can release gliotransmitters, which are essential for synaptic plasticity. Various mechanisms have been proposed for gliotransmission, including vesicular mechanisms as well as non-vesicular ones, for example by passive diffusion via
Wei Zhang et al.
Behavioral and brain functions : BBF, 19(1), 23-23 (2023-12-19)
As a kind of environmental noise, infrasonic noise has negative effects on various human organs. To date, research has shown that infrasound impairs cognitive function, especially the ability for learning and memory. Previously, we demonstrated that impaired learning and memory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service