Skip to Content
Merck
All Photos(1)

Key Documents

SML2304

Sigma-Aldrich

Clozapine N-oxide hydrochloride

≥98% (HPLC), powder, muscarinic DREADD activator

Synonym(s):

Clozapine N-oxide hydrochloride, 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e](1,4)diazepine N-oxide, hydrochloride salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4O · xHCl
CAS Number:
Molecular Weight:
342.82 (free base basis)
UNSPSC Code:
12352119
NACRES:
NA.77

Product Name

Clozapine N-oxide hydrochloride, ≥98% (HPLC), Water soluble Clozapine N-oxide

biological source

synthetic

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

solubility

DMSO: 40 mg/mL
water: 40 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=CC2=C(NC(C=CC=C3)=C3C(N4CC[N+]([O-])(C)CC4)=N2)C=C1.[xHCl]

InChI

1S/C18H19ClN4O/c1-23(24)10-8-22(9-11-23)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

OGUCZBIQSYYWEF-UHFFFAOYSA-N

General description

Clozapine N-oxide hydrochloride is a water soluble form of Clozapine N-oxide (CNO) (Sigma cat# C0832). CNO is a pharmacologically inert metabolite of the atypical anti-psychotic drug clozapine. CNO is a ligand for the engineered Gq protein coupled receptors (GPCRs) in the chemogenetic Designer Receptors Exclusively Activated by Designer Drug (DREADD). The typical use of DREADD is testing, enhancing neuronal firing and activating Gq signaling in neuronal and non-neuronal cells.

The selective targeting of DREADD systems to cell population can be achieved by using a cell type-specific promoter to drive DREADD expression, and the expression of this promoter can be further controlled using a recombinase-based system. DREADDs possess a low affinity for endogenous ligands and little constitutive activity, but may be activated by synthetic compounds such as CNO.

DREADD systems are activated by low nM concentrations of CNO and mobilize intracellular calcium. CNO appears to be pharmacologically and behaviorally inert in mice and rats when administered at the recommended doses (generally 0.1–3 mg/kg). CNO is commonly administrated via injection but also can be mixed into food chow or drinking water.

Other Notes

This is not a pharmaceutical grade product.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Blaine N Armbruster et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(12), 5163-5168 (2007-03-16)
We evolved muscarinic receptors in yeast to generate a family of G protein-coupled receptors (GPCRs) that are activated solely by a pharmacologically inert drug-like and bioavailable compound (clozapine-N-oxide). Subsequent screening in human cell lines facilitated the creation of a family
Masaki Ishikawa et al.
The FEBS journal, 291(5), 945-964 (2023-12-01)
Indoleamine 2,3-dioxygenase 2 (IDO2) is an enzyme of the tryptophan-kynurenine pathway that is constitutively expressed in the brain. To provide insight into the physiological role of IDO2 in the brain, behavioral and neurochemical analyses in IDO2 knockout (KO) mice were
Susan M Ferguson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(28), 11668-11676 (2013-07-12)
The dorsal striatum has been implicated in reward-based decision making, but the role played by specific striatal circuits in these processes is essentially unknown. Using cell phenotype-specific viral vectors to express engineered G-protein-coupled DREADD (designer receptors exclusively activated by designer
Daniel J Urban et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 41(5), 1404-1415 (2015-09-19)
Elucidating how the brain's serotonergic network mediates diverse behavioral actions over both relatively short (minutes-hours) and long period of time (days-weeks) remains a major challenge for neuroscience. Our relative ignorance is largely due to the lack of technologies with robustness
Hu Zhu et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 1880-1892 (2014-02-15)
Synaptic consolidation is a process thought to consolidate memory in the brain. Although lesion studies have mainly implicated the hippocampus (HPC) in this process, it is unknown which cell type(s) or regions of the HPC might be essential for synaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service