Skip to Content
Merck
All Photos(1)

Key Documents

SML2122

Sigma-Aldrich

ELND006

≥98% (HPLC)

Synonym(s):

(R)-4-Cyclopropyl-7,8-difluoro-5-(4-(trifluoromethyl)phenylsulfonyl)-4,5-dihydro-1H-pyrazolo[4,3-c]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14F5N3O2S
CAS Number:
Molecular Weight:
455.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -125 to -140°, c = 0.5 in ethanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ELND006 is an allosteric γ-secretase inhibitor that selectively blocks amyloid-β (Aβ) over Notch production both in cell-free (Aβ IC50/Notch IC50 = 0.34 nM/6.6 nM) and cell-based assays (Aβ IC50/Notch IC50 = 1.3 nM/85 nM). ELND006 is orally available and blood-brain barrier-permeable (total/unbound conc in brain = 0.12 μM/1.0 nM, 0.52 μM/4.5 nM, 2.1 μM/18 nM and total/unbound conc in plasma = 0.031 μM/0.28 nM, 0.19 μM/1.7 nM, 0.90 μM/8.1 nM 3 h post 1, 3, 10 mg/kg oral dosage in rats, respectively), and shown to effectively reduce cortical Aβx-40 in mice in vivo (by 32%, 48%, and 61% 3 h post 1, 3, 10 mg/kg oral dosage).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inger Lauritzen et al.
Acta neuropathologica, 132(2), 257-276 (2016-05-04)
Endosomal-autophagic-lysosomal (EAL) dysfunction is an early and prominent neuropathological feature of Alzheimers's disease, yet the exact molecular mechanisms contributing to this pathology remain undefined. By combined biochemical, immunohistochemical and ultrastructural approaches, we demonstrate a link between EAL pathology and the
Fiorenza Stagni et al.
Neurobiology of disease, 103, 11-23 (2017-04-01)
Neurogenesis impairment is considered a major determinant of the intellectual disability that characterizes Down syndrome (DS), a genetic condition caused by triplication of chromosome 21. Previous evidence obtained in the Ts65Dn mouse model of DS showed that the triplicated gene
Gary Probst et al.
Journal of medicinal chemistry, 56(13), 5261-5274 (2013-05-30)
Herein, we describe our strategy to design metabolically stable γ-secretase inhibitors which are selective for inhibition of Aβ generation over Notch. We highlight our synthetic strategy to incorporate diversity and chirality. Compounds 30 (ELND006) and 34 (ELND007) both entered human
Rampurna Gullapalli et al.
Drug delivery, 19(5), 239-246 (2012-06-05)
Hydrophilic, non-aqueous solvents are frequently used to solubilize poorly water soluble compounds for use in ALZET® osmotic pumps used during the discovery and preclinical stages. Though these solvents exhibit the potential to solubilize several poorly soluble compounds, the solubilized compounds
Dolores Del Prete et al.
Journal of Alzheimer's disease : JAD, 55(4), 1549-1570 (2016-12-03)
Alteration of mitochondria-associated membranes (MAMs) has been proposed to contribute to the pathogenesis of Alzheimer's disease (AD). We studied herein the subcellular distribution, the processing, and the protein interactome of the amyloid-β protein precursor (AβPP) and its proteolytic products in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service