Skip to Content
Merck
All Photos(1)

Key Documents

SML2099

Sigma-Aldrich

NIBR-0213

≥98% (HPLC)

Synonym(s):

(S)-2-({3′-[(R)-1-(4-Chloro-3-methyl-phenyl)-ethylamino]-3,5-dimethyl-biphenyl-4-carbonyl}-amino)-propionic acid, N-[[3′-[[(1R)-1-(4-Chloro-3-methylphenyl)ethyl]amino]-3,5-dimethyl[1,1′-biphenyl]-4-yl]carbonyl]-L-alanine, NIBR 0213, NIBR0213

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29ClN2O3
CAS Number:
Molecular Weight:
464.98
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@@H](C(O)=O)NC(C1=C(C)C=C(C2=CC=CC(N[C@@H](C3=CC=C(Cl)C(C)=C3)C)=C2)C=C1C)=O

InChI

1S/C27H29ClN2O3/c1-15-11-20(9-10-24(15)28)18(4)29-23-8-6-7-21(14-23)22-12-16(2)25(17(3)13-22)26(31)30-19(5)27(32)33/h6-14,18-19,29H,1-5H3,(H,30,31)(H,32,33)/t18-,19+/m1/s1

InChI key

KYHUARFFBDLROH-MOPGFXCFSA-N

Application

NIBR-0213 may be used as a selective sphingosine-1-phosphate receptor 1 (S1P1) antagonist to assess S1P-mediated nuclear factor kappa B (NF-κB)-p65 nuclear translocation in human induced astrocytes.

Biochem/physiol Actions

NIBR-0213 is an orally active, potent and S1P1-selective sphingosine 1-phosphate (S1P) receptor antagonist that blocks S1P1-selective ligand AUY954-induced Ca2+ mobilization (IC50 = 2.5 nM using human S1P1-transfected HeLa cells; IC50 >10 μM with human S1P2, S1P3, or S1P4 transfectants) and S1P-induced GTPγS recruitment (IC50 = 2.0/2.3/8.5 nM using membrane from human/murine/rat S1P1-transfected CHO cells; IC50 >10 μM with membrane from human S1P5 transfectant). NIBR-0213 induces long-lasting peripheral blood lymphocyte reduction in rats (EDmax = 1 mg/kg, ED50 = 0.2 mg/kg; p.o.) and exhibits therapeutic efficacy (30 mg/kg p.o; BID) in experimental autoimmune encephalomyelitis (EAE), a murine model of multiple sclerosis (MS), with no adverse effects to the animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Colombo et al.
Frontiers in immunology, 11, 635-635 (2020-04-24)
Multiple sclerosis (MS) is an inflammatory neurodegenerative disease of the central nervous system (CNS) with heterogeneous pathophysiology. In its progressive course oligodendrocyte and neuroaxonal damage is sustained by compartmentalized inflammation due to glial dysregulation. Siponimod (BAF312), a modulator of two
Marc Bigaud et al.
PloS one, 11(12), e0168252-e0168252 (2016-12-23)
Homeostasis of vascular barriers depends upon sphingosine 1-phosphate (S1P) signaling via the S1P1 receptor. Accordingly, S1P1 competitive antagonism is known to reduce vascular barrier integrity with still unclear pathophysiological consequences. This was explored in the present study using NIBR-0213, a
Victoria Fang et al.
Nature immunology, 18(1), 15-25 (2016-11-15)
The lymph node periphery is an important site for many immunological functions, from pathogen containment to the differentiation of helper T cells, yet the cues that position cells in this region are largely undefined. Here, through the use of a
Jean Quancard et al.
Chemistry & biology, 19(9), 1142-1151 (2012-09-25)
Lymphocyte trafficking is critically regulated by the Sphingosine 1-phosphate receptor-1 (S1P(1)), a G protein-coupled receptor that has been highlighted as a promising therapeutic target in autoimmunity. Fingolimod (FTY720, Gilenya) is a S1P(1) receptor agonist that has recently been approved for
Keisuke Yanagida et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(17), 4531-4536 (2017-04-12)
The vasculature of the central nervous system (CNS) forms a selective barrier termed the blood-brain barrier (BBB). Disruption of the BBB may contribute to various CNS diseases. Conversely, the intact BBB restricts efficient penetration of CNS-targeted drugs. Here, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service