Skip to Content
Merck
All Photos(1)

Key Documents

SML1264

Sigma-Aldrich

Rosuvastatin calcium

≥98% (HPLC)

Synonym(s):

(E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrim idin-5-yl]-(3R,5S)-3,5-dihydroxyhept-6-enoic acid, calcium salt (2:1), Rosuvastatin hemicalcium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27FN3O6S · 0.5 Ca
CAS Number:
Molecular Weight:
500.57
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +12 to +18°, c = 1 in methanol: water (1:1)

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Ca+2].Fc3ccc(cc3)c4nc(nc(c4\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])C(C)C)N([S](=O)(=O)C)C.Fc1ccc(cc1)c2nc(nc(c2\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])C(C)C)N([S](=O)(=O)C)C

InChI

1S/2C22H28FN3O6S.Ca/c2*1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h2*5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;;+2/p-2/b2*10-9+;/t2*16-,17-;/m11./s1

InChI key

LALFOYNTGMUKGG-BGRFNVSISA-L

General description

Rosuvastatin calcium (ROS) has a molecular mass of 1001.14. ROS is a member of the "statins" group and is a 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor.

Application

Rosuvastatin calcium has been used to determine the effect of the treatment with SIM, rosuvastatin or PRA on cholesterol and HMGCR levels in HeLa cells and to evaluate the in vivo effects of statins on glucose homeostasis.

Biochem/physiol Actions

Rosuvastatin calcium is a HMG-CoA reductase inhibitor.
Rosuvastatin calcium is a competitive inhibitor of hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase, the enzyme that catalyzes the conversion of HMG-CoA to mevalonic acid, the rate-limiting step in cholesterol biosynthesis. Rosuvastatin calcium is antilipemic and is used to reduce plasma cholesterol levels and prevent cardiovascular disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholesterol crystal formation is a unifying pathogenic mechanism in the development of diabetic retinopathy.
Hammer, et al.
Diabetologia (2023)
Analytical Methods for the Determination of Rosuvastatin in Pharmaceutical Formulations and Biological Fluids: A Critical Review.
Angelo ML, et al.
Critical Reviews in Analytical Chemistry, 48(4), 317-329 (2018)
HPLC methods for recently approved pharmaceuticals (2005)
Chronic HMGCR/HMG-CoA reductase inhibitor treatment contributes to dysglycemia by upregulating hepatic gluconeogenesis through autophagy induction.
Wang HJ, et al.
Autophagy, 11(11), 2089-2101 (2015)
Joel P Wiener et al.
Cells, 12(17) (2023-09-08)
Adenosine triphosphate-binding cassette transporter subfamily A member 7 (ABCA7) is a major risk factor for Alzheimer's disease. Human neural cell lines were used to investigate the regulation of ABCA7 expression by cholesterol and pro-inflammatory cytokines. Cholesterol was depleted by methyl-β-cyclodextrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service