Skip to Content
Merck

SML1002

Sinomenine hydrochloride

≥98% (HPLC)

Synonym(s):

(9a,13a,14a)-7,8-Didehydro-4-hydroxy-3,7-dimethoxy-17-methyl-morphinan-6-one hydrochloride, Cucoline hydrochloride, Kukolin hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H23NO4 · HCl
CAS Number:
Molecular Weight:
365.85
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant (Sinomenium acutum)

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -70 to -90°, c = 1 in H2O

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(OC)C=CC(C2)=C1[C@]3(CC(C(OC)=C4)=O)[C@@]4([H])[C@H]2N(C)CC3.Cl

InChI

1S/C19H23NO4.ClH/c1-20-7-6-19-10-14(21)16(24-3)9-12(19)13(20)8-11-4-5-15(23-2)18(22)17(11)19;/h4-5,9,12-13,22H,6-8,10H2,1-3H3;1H/t12-,13+,19-;/m1./s1

InChI key

YMEVIMJAUHZFMW-VUIDNZEBSA-N

Biochem/physiol Actions

Sinomenine is a herbal medicine used to treat inflammatory diseases like rheumatoid arthritis. Sinomenine had anti-inflammatory effects in several animal models of inflammation, and inhibits mast cell degranulation and histamine release. Additionally, Sinomenine potentiates renewal of human embryonic stem cells.
Sinomenine is a herbal medicine used to treat inflammatory diseases.


Still not finding the right product?

Explore all of our products under Sinomenine hydrochloride


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library