Skip to Content
Merck

Skip To

SML0940

Nepicastat hydrochloride

≥95% (HPLC)

Synonym(s):

5-(Aminomethyl)-1-[(2S)-5,7-difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]-1,3-dihydro-2H-imidazole-2-thione hydrochloride, SYN-117

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H15F2N3S · HCl
CAS Number:
Molecular Weight:
331.81
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=CC2=C(CC[C@H](N3C(CN)=CNC3=S)C2)C(F)=C1.Cl

InChI

1S/C14H15F2N3S.ClH/c15-9-3-8-4-10(1-2-12(8)13(16)5-9)19-11(6-17)7-18-14(19)20;/h3,5,7,10H,1-2,4,6,17H2,(H,18,20);1H/t10-;/m0./s1

InChI key

DIPDUAJWNBEVOY-PPHPATTJSA-N

Application

Nepicastat hydrochloride has been used as an inhibitor of dopamine β-hydroxylase (DBH) in zinc finger protein 521 (ZFP521) deficient mice.

Biochem/physiol Actions

Nepicastat is a potent and selective dopamine β-hydroxylase inhibitor
Nepicastat is a potent and selective dopamine β-hydroxylase inhibitor with an IC50 of 9 nM and negligible affinity (>10 μM) for twelve other enzymes and thirteen neurotransmitter receptors tested. Nepicastat has been shown to attenuate multiple aspects of cocaine-seeking behavior and suppress cocaine-primed reinstatement of cocaine seeking in rats, and has been in clinical trials for cocaine dependence. Nepicastat also suppressed chocolate self-administration and reinstatement of chocolate seeking in rats.
By inhibiting dopamine β-hydroxylase (DβH), nepicastat enhances dopamine levels and norepinephrine levels in brain.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0990SML0787SML0893
description

≥95% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

desiccated

storage condition

-

storage condition

desiccated

storage condition

desiccated

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

H2O: 2 mg/mL, clear (warmed)


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters