Skip to Content
Merck
All Photos(1)

Key Documents

SML0822

Sigma-Aldrich

YM-08

≥98% (HPLC)

Synonym(s):

3-Ethyl-5-(3-methyl-2(3H)-benzothiazolylidene)-2-(2-pyridinylmethylene)-4-thiazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3OS2
CAS Number:
Molecular Weight:
367.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

storage temp.

2-8°C

Biochem/physiol Actions

YM-08 is a Heat Shock Protein 70 (Hsp70) inhibitor that is blood-brain barrier permeable. YM-08 is a neutral derivative of the Hsp70 inhibitor MKT-077. MKT-077 reduces tau levels in cellular models, but it doesn′t penetrate the blood brain barrier. Although YM-08 was somewhat less effective than MKT-077 in anti-tau and anticancer assays, it selectively reduced pathogenic tau in brain slices, which MKT-077 did not. Furthermore,YM-08 was not retained in the kidney leading one to speculate that it would not have the nephrotoxicity that was a problem for MKT-077 in clinical trials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service