Skip to Content
Merck
All Photos(1)

Key Documents

SML0717

Sigma-Aldrich

W-13

≥98% (HPLC)

Synonym(s):

N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide hydrochloride, W13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17ClN2O2S · HCl
CAS Number:
Molecular Weight:
349.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

Cl.NCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12

InChI

1S/C14H17ClN2O2S.ClH/c15-13-7-3-6-12-11(13)5-4-8-14(12)20(18,19)17-10-2-1-9-16;/h3-8,17H,1-2,9-10,16H2;1H

InChI key

IKMXJMNRHREPOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

W-13 has also been shown to inhibit the transcytosis of IgA, recycling of transferrin and overall alter the endocytic pathway in Madin-Darby canine kidney cells.
W-13 is a membrane-permeable calmodulin antagonist. W-13 inhibits Ca2+-calmodulin-dependent phosphodiesterase and has been shown to inhibit the EGF-dependent activation of EGFR in N7xHERc fibroblasts.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Speranza et al.
Food microbiology, 87, 103393-103393 (2020-01-18)
Four wild strains of Saccharomyces cerevisiae and the collection strain S. cerevisiae var. boulardii ATCC MYA-796 were used as test organisms to study the effect of some environmental conditions on the formation of biofilm by potentially probiotic yeasts. In a
Danying Zhou et al.
Frontiers in microbiology, 11, 2021-2021 (2020-09-26)
Yokenella regensburgei, a member of the family Enterobacteriaceae, is usually isolated from environmental samples and generally resistant to early generations of cephalosporins. To characterize the resistance mechanism of Y. regensburgei strain W13 isolated from the sewage of an animal farm
Albert Lu et al.
The Journal of cell biology, 184(6), 863-879 (2009-03-18)
Ras proteins are small guanosine triphosphatases involved in the regulation of important cellular functions such as proliferation, differentiation, and apoptosis. Understanding the intracellular trafficking of Ras proteins is crucial to identify novel Ras signaling platforms. In this study, we report
The calmodulin antagonist, W-13, alters transcytosis, recycling, and the morphology of the endocytic pathway in Madin-Darby canine kidney cells.
Apodaca G
The Journal of Biological Chemistry (1994)
Antonio Bevilacqua et al.
Foods (Basel, Switzerland), 9(8) (2020-08-08)
Five yeast strains (four wild Saccharomyces cerevisiae strains and a collection strain-S. cerevisiae var. boulardii) were encapsulated in alginate beads. Encapsulation yield was at least 60% (100% for some strains) and yeasts survived in beads for 30 days at 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service