Skip to Content
Merck
All Photos(1)

Key Documents

SML0462

Sigma-Aldrich

Alda-1

≥98% (HPLC)

Synonym(s):

N-(1,3-Benzodioxol-5-ylmethyl)-2,6-dichlorobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl2NO3
CAS Number:
Molecular Weight:
324.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Clc1cccc(Cl)c1C(=O)NCc2ccc3OCOc3c2

InChI

1S/C15H11Cl2NO3/c16-10-2-1-3-11(17)14(10)15(19)18-7-9-4-5-12-13(6-9)21-8-20-12/h1-6H,7-8H2,(H,18,19)

InChI key

NMKJFZCBCIUYHI-UHFFFAOYSA-N

Application

Alda-1 has been used as an aldehyde dehydrogenase 2 (ALDH2) agonist:
  • to study its cryoprotective effects on H2O2-induced Achilles tendinopathy in mice Achilles tenocytes
  • to study its protective effects against alcohol-derived DNA damage in ALDH2 knock out mice
  • to study its effects on cutaneous wound healing in mice

Biochem/physiol Actions

Alda-1 is an ALDH2 agonist; cell-permeable activator of both the wild-type ALDH2*1 and the asian E487K mutant ALDH2*2 forms of mitochondrial aldehyde dehydrogenase 2.
Alda-1 is an ALDH2 agonist; cell-permeable activator of both the wild-type ALDH2*1 and the asian E487K mutant ALDH2*2 forms of mitochondrial aldehyde dehydrogenase 2. ALDH2 is the oxidative enzyme, which removes the ethanol metabolite acetaldehyde and other aliphatic aldehydes. Also. ALDH2 is involved in bioconversion of vasodilator nitroglycerin to nitric oxide. Alda-1 increases acetaldehyde oxidation by ALDH2*1 (wild type) and ALDH2*2 9 Asian variant) approximately 1.5- and 6-fold, respectively. It appears that Alda-1 stimulates acetaldehyde oxidation by ALDH2 by improving NAD binding, but does not improve the nitroglycerin binding affinity of the Asian variant.
Alda-1 plays a role in reducing the damage caused by oxidative stress in several human cells like lung endothelial, umbilical vein endothelial, and hepatic cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tingting Fang et al.
Molecular medicine reports, 18(3), 2807-2815 (2018-07-18)
The aim of the present study was to investigate whether necroptosis occurs in high glucose (HG)-induced H9c2 cardiac cell injury and whether the activation of aldehyde dehydrogenase 2 (ALDH2) can inhibit necroptosis. H9c2 cardiac cells were treated with 35 mM glucose
Gina D Kusuma et al.
Scientific reports, 7, 42397-42397 (2017-02-17)
High resistance to oxidative stress is a common feature of mesenchymal stem/stromal cells (MSC) and is associated with higher cell survival and ability to respond to oxidative damage. Aldehyde dehydrogenase (ALDH) activity is a candidate "universal" marker for stem cells.
Jianlu Guo et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 38(11), 1288-1293 (2018-12-06)
To investigate whether CaN-NFAT3 pathway mediates the protective effects of aldehyde dehydrogenase (ALDH) 2 in high glucose-treated neonatal rat ventricular myocytes. The ventricular myocytes were isolated from the heart of neonatal (within 3 days) SD rats by enzyme digestion and
Fayez T Hammad et al.
Drug development research, 79(7), 315-323 (2018-10-07)
Renal ischemia-reperfusion injury (IRI) induces the production of aldehydes which are detoxified by aldehyde dehydrogenases (ALDHs). Alda-1 is a selective ALDH2 agonist and its protective effect was demonstrated in several conditions. The effect of Alda-1 on the kidney or on
Qiankun Zhu et al.
Clinical science (London, England : 1979), 131(11), 1123-1136 (2017-03-23)
Many studies demonstrate that activation of aldehyde dehydrogenase 2 (ALDH2) protects against oxidative stress via detoxification of cytotoxic aldehydes, and could attenuate cardiac, cerebral, lung and renal ischaemia-reperfusion (I/R) injuries. However, the effect of ALDH2 in intestinal I/R is unknown. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service