Skip to Content
Merck
All Photos(1)

Key Documents

SML0356

Sigma-Aldrich

Etonogestrel

≥98% (HPLC)

Synonym(s):

(17α)-13-Ethyl-17-hydroxy-11-methylene-18,19-dinorpregn-4-en-20-yn-3-one, 3-Keto-desogestrel, 3-Oxo-desogestrel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O2
CAS Number:
Molecular Weight:
324.46
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +80 to +95°, c = 1 (CHCl3)

color

white to beige

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

InChI

1S/C22H28O2/c1-4-21-13-14(3)20-17-9-7-16(23)12-15(17)6-8-18(20)19(21)10-11-22(21,24)5-2/h2,12,17-20,24H,3-4,6-11,13H2,1H3/t17-,18-,19-,20+,21-,22-/m0/s1

InChI key

GCKFUYQCUCGESZ-BPIQYHPVSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Etonogestrel is a lipophilic molecule. Pharmacokinetics studies reveal that etonogestrel is bound to protein albumin in the blood. This remains independent of both endogenous and exogenous concentration of estradiol.
Etonogestrel is a progestin, contraceptive.
Etonogestrel is a third-generation progestin contraceptive. Etonogestrel is used as a contraceptive itself and is also the active metabolite of the drug desogestrel used in some combination contraceptives.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anne Calhoun et al.
Headache, 52(8), 1246-1253 (2012-07-14)
To determine whether extended-cycle dosing of an ultralow dose vaginal ring contraceptive decreases frequency of migraine aura and prevents menstrual related migraine (MRM). Many women are denied therapy with combined hormonal contraceptives due to published guidelines that recommend against their
Peter Schnabel et al.
Clinical drug investigation, 32(6), 413-422 (2012-05-01)
The etonogestrel (ENG)-releasing implant is a subdermal progestogen-only contraceptive that provides coverage for up to 3 years. This long-acting hormonal contraceptive has been available in Europe since 1998 and in the US since 2006. To date, localization of non-palpable implants
Pharmacokinetics of etonogestrel released from the contraceptive implant Implanon
Wenzl R, et al.
Contraception, 58(5), 283-288 (1998)
Diana Mansour et al.
Contraception, 83(3), 202-210 (2011-02-12)
The aim of this guidance is to review the management of unacceptable vaginal bleeding patterns in etonogestrel (ENG)-releasing contraceptive implant users concentrating, where possible, on the evidence for pharmacological treatments and identifying a pragmatic approach where this is not possible.
Hanna Xu et al.
Obstetrics and gynecology, 120(1), 21-26 (2012-06-09)
To estimate the contraceptive failure rates of the etonogestrel subdermal contraceptive implant in overweight and obese women and compare failure rates with women of normal weight and women using intrauterine devices (IUDs). The Contraceptive CHOICE Project is a large prospective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service