Skip to Content
Merck
All Photos(1)

Key Documents

SML0326

Sigma-Aldrich

GTS-21

≥97% (HPLC)

Synonym(s):

3-(2,4-Dimethoxybenzylidene)-anabaseine dihydrochloride, DMBX-anabaseine, DMXB, DMXB-A, GTS21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O2 · 2HCl
CAS Number:
Molecular Weight:
381.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Cl.COc1ccc(\C=C2/CCCN=C2c3cccnc3)c(OC)c1

InChI

1S/C19H20N2O2.2ClH/c1-22-17-8-7-14(18(12-17)23-2)11-15-5-4-10-21-19(15)16-6-3-9-20-13-16;;/h3,6-9,11-13H,4-5,10H2,1-2H3;2*1H/b15-11+;;

InChI key

BXKYFUGAAFLYJL-BXGYHSFXSA-N

Application

GTS-21 has been used:
  • as an α7 nicotinic acetylcholine receptors (nAChR) partial agonist to elucidate its anti-inflammatory effects in mouse macrophages
  • to test its protective effect on the renal injury induced by lipopolysaccharide (LPS)
  • to test its effect on microvascular inflammation in endotoxemia induced by LPS

Biochem/physiol Actions

GTS-2, a derivative of anisine is an immunomodulatory drug. It is used for treating pancreatitis and septicemia. GTS-2 inhibits the pro-inflammatory cytokines especially the interleukin-6 (IL6) and tumor necrosis factor (TNF) in sepsis and endotoxemia.
GTS-21 is a selective agonist at α-7 nicotinic receptors with anti-inflammatory and cognition enhancing activities. GTS-21 has also been investigated for the treatment of schizophrenia.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca Brawek et al.
International journal of molecular sciences, 22(3) (2021-01-28)
Microglia, the innate immune cells of the brain, are commonly perceived as resident macrophages of the central nervous system (CNS). This definition, however, requires further specification, as under healthy homeostatic conditions, neither morphological nor functional properties of microglia mirror those
Ryan E Hibbs et al.
The EMBO journal, 28(19), 3040-3051 (2009-08-22)
The pentameric acetylcholine-binding protein (AChBP) is a soluble surrogate of the ligand binding domain of nicotinic acetylcholine receptors. Agonists bind within a nest of aromatic side chains contributed by loops C and F on opposing faces of each subunit interface.
Caijuan Shi et al.
Journal of molecular modeling, 19(2), 871-878 (2012-10-23)
Nicotinic acetylcholine receptors (nAChRs) are drug targets for neuronal disorders and diseases. Partial agonists for nAChRs are currently being developed as drugs for the treatment of neurological diseases for their relative safety originated from reduced excessive stimulation. In the current
Prodyot K Chatterjee et al.
PloS one, 7(5), e35361-e35361 (2012-05-16)
Sepsis is one of the leading causes of acute kidney injury (AKI). Septic patients who develop acute kidney injury (AKI) are at increased risk of death. To date there is no effective treatment for AKI or septic AKI. Based on
Jerry J Buccafusco et al.
Biochemical pharmacology, 78(7), 852-862 (2009-07-07)
In monkeys proficient in the performance of a computer-assisted delayed response task, administration of sub-sedative doses of ketamine significantly impaired task performance after the 2mg/kg dose, producing a decrease in accuracies across all four delay intervals. Ketamine elicited occasional and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service