Skip to Content
Merck
All Photos(1)

Key Documents

SML0308

Sigma-Aldrich

Carbazeran

≥95% (HPLC)

Synonym(s):

N-Ethyl-carbamic acid 1-(6,7-dimethoxy-1-phthalazinyl)-4-piperidinyl ester, UK 31557

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N4O4
CAS Number:
Molecular Weight:
360.41
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥2 mg/mL at warmed

storage temp.

2-8°C

SMILES string

CCNC(=O)OC1CCN(CC1)c2nncc3cc(OC)c(OC)cc23

InChI

1S/C18H24N4O4/c1-4-19-18(23)26-13-5-7-22(8-6-13)17-14-10-16(25-3)15(24-2)9-12(14)11-20-21-17/h9-11,13H,4-8H2,1-3H3,(H,19,23)

InChI key

QJGVXJYGDBSPSJ-UHFFFAOYSA-N

General description

Carbazeran is usedin the treatment of heart failure.

Biochem/physiol Actions

Carbazeran is an Aldehyde oxidase (AO) substrate and a phosphodiesterase inhibitor that produces concentration-dependent positive inotropic responses. In humans, the compound is almost completely cleared via 4-hydroxylation to the phthalazinone metabolite by AO.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C R Taylor et al.
American heart journal, 102(3 Pt 2), 515-532 (1981-09-01)
The animal and human pharmacology of several new drugs (prazosin, trimazosin, pirbuterol, and carbazeran) useful in the treatment of congestive heart failure (CHF) is delineated in relation to the pharmacology of other agents employed for CHF management. Prazosin and trimazosin
D el Allaf et al.
Archives internationales de physiologie et de biochimie, 92(4), S69-S79 (1984-11-01)
Compounds with phosphodiesterase inhibitory activity stimulate myocardial contractility by increasing the intracellular cyclic AMP concentrations. They can also increase Ca2+ entry and inhibit Ca2+ sequestration by the sarcoplasmic reticulum. Xanthines produce bronchodilation with associated venous and arteriolar dilation. However, their
Wee Kiat Tan et al.
The Journal of pharmacology and experimental therapeutics, 374(2), 295-307 (2020-05-13)
Gefitinib and erlotinib are epidermal growth factor receptor-tyrosine kinase inhibitors (EGFR-TKIs) with activity against metastatic non-small cell lung cancer. Aldehyde oxidase-1 (AOX1) is a cytosolic drug-metabolizing enzyme. We conducted an experimental and molecular docking study on the effect of gefitinib
Shotaro Uehara et al.
Drug metabolism and disposition: the biological fate of chemicals, 48(7), 580-586 (2020-05-03)
Carbazeran is a potent phosphodiesterase inhibitor with species-dependent metabolic profiles in rats, dogs, and humans. In this study, we investigated the aldehyde oxidase (AOX)-mediated oxidation of carbazeran to 4-oxo derivatives in chimeric NOD/Shi-scid IL2 receptor gamma-null mice expressing a herpes
B Kaye et al.
Xenobiotica; the fate of foreign compounds in biological systems, 14(12), 935-945 (1984-12-01)
The bioavailability of carbazeran and the metabolism of carbon-14 labelled drug have been studied in the dog and man following oral administration. The drug was moderately well absorbed in both species, but there was a marked difference in bioavailability and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service