Skip to Content
Merck
All Photos(1)

Documents

SML0289

Sigma-Aldrich

Bromfenac sodium

≥98% (HPLC)

Synonym(s):

2-Amino-3-(4-bromobenzoyl)benzeneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11BrNNaO3
CAS Number:
Molecular Weight:
356.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

faintly yellow to dark yellow

solubility

H2O: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

[Na+].Nc1c(CC([O-])=O)cccc1C(=O)c2ccc(Br)cc2

InChI

1S/C15H12BrNO3.Na/c16-11-6-4-9(5-7-11)15(20)12-3-1-2-10(14(12)17)8-13(18)19;/h1-7H,8,17H2,(H,18,19);/q;+1/p-1

InChI key

HZFGMQJYAFHESD-UHFFFAOYSA-M

Gene Information

Application

Bromfenac sodium has been used:
  • to study its ability to bind to melanin
  • in the synthesis of bromfenac indolinone standard
  • to analyze its permeability in porcine conjunctiva

Biochem/physiol Actions

Bromfenac exhibits antipyretic and prostaglandin synthetase inhibiting properties. It has therapeutic properties against the reduction of ocular pain and inflammation in postoperative cataract patients. Bromfenac acts as an effective agent against allergic conjunctivitis. It has the potential to treat acute muscle pain, osteoarthritis, and rheumatoid arthritis.
Bromfenac is a nonsteroidal anti inflammatory drug (NSAID) that inhibits both COX1 and COX2. It is used as an opthalmic analgesic.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Keith A Warren et al.
Retina (Philadelphia, Pa.), 30(2), 260-266 (2010-02-23)
The purpose of this study was to evaluate the addition of topical nonsteroidal antiinflammatory drugs (NSAIDs) to intravitreal corticosteroid and antivascular endothelial growth factor injections for the treatment of chronic cystoid macular edema. Thirty-nine patients with chronic pseudophakic cystoid macular
Ester Carreño et al.
Clinical ophthalmology (Auckland, N.Z.), 6, 637-644 (2012-05-10)
Ophthalmic bromfenac sodium sesquihydrate is a topically applied selective cyclooxygenase (COX)-2 inhibitor. It is similar to amfenac, except for a bromine atom at the C(4) of the benzoyl ring position, which markedly affects its in vitro and in vivo potency
Neal A Sher et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 25(2), 214-220 (2009-02-27)
To compare the efficacy of two topical nonsteroidal anti-inflammatory drugs with regards to the control of pain, burning, photophobia, foreign body sensation, and epithelial healing rates in patients who underwent photorefractive keratectomy (PRK). Two hundred twelve eyes were randomized to
Christina Flaxel et al.
Retina (Philadelphia, Pa.), 32(3), 417-423 (2011-08-25)
To evaluate whether bromfenac eyedrops and ranibizumab intravitreal injections would provide added efficacy over ranibizumab alone. This was a single-site, multiinvestigator, prospective, open-label, interventional, Phase II study of patients with new or recurrent exudative/neovascular age-related macular degeneration. Thirty eyes were
Frank A Bucci et al.
Advances in therapy, 28(12), 1089-1095 (2011-11-23)
We compared the prostaglandin E(2) (PGE(2)) inhibition of three topical nonsteroidal antiinflammatory drugs (NSAIDs): ketorolac 0.45%, bromfenac 0.09%, and nepafenac 0.1% at peak dosing levels in patients previously scheduled to undergo phacoemulsification. This was a single-center, double-masked observational study of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service