Skip to Content
Merck
All Photos(1)

Key Documents

SML0238

Sigma-Aldrich

WEB2086

≥98% (HPLC)

Synonym(s):

3-[4-(2-Chlorophenyl)-9-methyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-2-yl]-1-(4-morpholinyl)-1-propanone, Apafant, WEB 2086BS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22ClN5O2S
CAS Number:
Molecular Weight:
455.96
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Cc1nnc2CN=C(c3ccccc3Cl)c4cc(CCC(=O)N5CCOCC5)sc4-n12

InChI

1S/C22H22ClN5O2S/c1-14-25-26-19-13-24-21(16-4-2-3-5-18(16)23)17-12-15(31-22(17)28(14)19)6-7-20(29)27-8-10-30-11-9-27/h2-5,12H,6-11,13H2,1H3

InChI key

JGPJQFOROWSRRS-UHFFFAOYSA-N

Biochem/physiol Actions

WEB 2086 inhibits PAF mediated chemotaxis and leukotriene C4 production in eosinophils. It prevents surge in airway resistance and cardiovascular side effects mediated by PAF inhalation.
WEB2086 is a very potent, selective platelet-activating factor (PAF) antagonist (IC50 = 16 nM). WEB2086 inhibits the proliferation of tumor cells and slows tumor growth in xenograft tumor models, and has been shown to inhibit angiogenesis.
WEB2086 is a very potent, selective platelet-activating factor (PAF) antagonist.

Features and Benefits

This compound is featured on the PAF Receptor page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evandro M M Machado et al.
Journal of insect physiology, 52(7), 685-692 (2006-06-17)
The effects of the triazolodiazepine WEB 2086, a platelet-activating factor (PAF) antagonist, on hemocyte microaggregation and prophenoloxidase (proPO)-activating system in the hemolymph, hemocoelic infection and mortality in fifth-instar larvae of Rhodnius prolixus inoculated with Trypanosoma rangeli were investigated. Hemocoelic injection
Inhibitory effects of the new PAF acether antagonist WEB-2086 on pharmacologic changes induced by PAF inhalation in human beings
Adamus WS, et al.
Clinical Pharmacology, 47(4), 456-462 (1990)
Eduardo Tibiriçá et al.
Journal of ethnopharmacology, 111(1), 50-55 (2006-12-08)
We investigated the vascular effects of a crude aqueous extract (AEEG) of Echinodorus grandiflorus (Alismataceae) using the in vitro experimental models of the rabbit isolated aorta and perfused kidney. Echinodorus grandiflorus, a native semi-aquatic plant widely distributed in Brazil, has
Valérie Declerck et al.
The Journal of organic chemistry, 74(5), 2004-2007 (2009-02-10)
The 1,3-dipolar cycloaddition of linear azido alkynes derived from protected beta-amino esters proceeds via diastereomeric differentiation to provide trans-disubstituted triazolodiazepines in good yields.
Izumi Kikuchi et al.
American journal of respiratory cell and molecular biology, 34(6), 760-765 (2006-02-04)
Neutrophilic inflammation observed with severe asthma is often associated with interleukin-8 (IL-8). Neutrophils can secrete a variety of mediators that may augment the migration of eosinophils. We have reported a positive correlation between the concentrations of neutrophils and eosinophils in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service