Skip to Content
Merck
All Photos(1)

Documents

SML0180

Sigma-Aldrich

Aceclidine hydrochloride

≥98% (HPLC)

Synonym(s):

1-Azabicyclo[2.2.2]oct-3-yl acetate hydrochloride, 3-Acetoxyquinuclidine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO2 · HCl
CAS Number:
Molecular Weight:
205.68
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥15 mg/mL at ~60 °C

storage temp.

−20°C

SMILES string

Cl.CC(=O)O[C@H]1CN2CC[C@H]1CC2

InChI

1S/C9H15NO2.ClH/c1-7(11)12-9-6-10-4-2-8(9)3-5-10;/h8-9H,2-6H2,1H3;1H/t9-;/m0./s1

InChI key

LWWSARSTZGNKGV-FVGYRXGTSA-N

Application

Aceclidine hydrochloride may be used in muscarinic acetylcholine receptor-mediated cell signaling studies.

Biochem/physiol Actions

Aceclidine at a concentration of 5 and 50 μg is capable of enhancing the outflow facility, following ciliary muscle disinsertion.
Aceclidine hydrochloride is a parasympathomimetic drug with weak anticholinesterase activity. It acts on cholinergic nerve-endings, lowers intraocular pressure and potent therapeutic agent for open-angle glaucoma.
Aceclidine is a non-selective muscarinic acetylcholine receptor partial agonist that is effective in reducing intraocular pressure.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aceclidine effects on outflow facility after ciliary muscle disinsertion
Hubbard WC, et al.
Ophthalmologica, 210(5), 303-307 (1996)
Double-blind cross-over comparison of aceclidine and pilocarpine in open-angle glaucoma.
J H Romano
The British journal of ophthalmology, 54(8), 510-521 (1970-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service