Skip to Content
Merck
All Photos(1)

Key Documents

SML0108

Sigma-Aldrich

R18 trifluoroacetate

≥98% (HPLC)

Synonym(s):

L-Prolyl-L-histidyl-L-cysteinyl-L-valyl-L-prolyl-L-arginyl-L-a-aspartyl-L-leucyl-L-seryl-L-tryptophyl-L-leucyl-L-a-aspartyl-L-leucyl-L-a-glutamyl-L-alanyl-L-asparaginyl-L-methionyl-L-cysteinyl-L-leucyl-L-proline trifluoroacetate, PHCVPRDLSWLDLEANMCLP trifluoroacetate, Pro-His-Cys-Val-Pro-Arg-Asp-Leu-Ser-Trp-Leu-Asp-Leu-Glu-Ala-Asn-Met-Cys-Leu-Pro trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C101H157N27O29S3
CAS Number:
Molecular Weight:
2309.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CSCC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]3CCCN3C(=O)[C@@H](NC(=O)[C@H](CS)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@@H]5CCCN5)C(C)C)C(=O)N[C@@H](CS)C(=O)N[C@@H](CC(C)C)C(=O)N6CCC[C@H]6C(O)=O

InChI

1S/C101H157N27O29S3.C2HF3O2/c1-48(2)33-62(86(142)111-60(25-26-77(131)132)83(139)110-53(11)81(137)114-67(39-76(102)130)91(147)112-61(27-32-160-12)85(141)124-72(45-158)95(151)122-70(36-51(7)8)98(154)128-31-18-24-75(128)100(156)157)116-93(149)69(41-79(135)136)121-87(143)63(34-49(3)4)115-89(145)65(37-54-42-108-57-20-14-13-19-56(54)57)118-94(150)71(44-129)123-88(144)64(35-50(5)6)117-92(148)68(40-78(133)134)120-84(140)59(22-16-29-107-101(103)104)113-97(153)74-23-17-30-127(74)99(155)80(52(9)10)126-96(152)73(46-159)125-90(146)66(38-55-43-105-47-109-55)119-82(138)58-21-15-28-106-58;3-2(4,5)1(6)7/h13-14,19-20,42-43,47-53,58-75,80,106,108,129,158-159H,15-18,21-41,44-46H2,1-12H3,(H2,102,130)(H,105,109)(H,110,139)(H,111,142)(H,112,147)(H,113,153)(H,114,137)(H,115,145)(H,116,149)(H,117,148)(H,118,150)(H,119,138)(H,120,140)(H,121,143)(H,122,151)(H,123,144)(H,124,141)(H,125,146)(H,126,152)(H,131,132)(H,133,134)(H,135,136)(H,156,157)(H4,103,104,107);(H,6,7)/t53-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,80-;/m0./s1

InChI key

RLMYFUMCZYJMNX-KIXMOXNVSA-N

General description

R18 trifluoroacetate is a nonphosphorylated peptide. This peptide was originally identified as a high-affinity 14-3-3 ligand by phage display.

Application

R18 trifluoroacetate has been used to treat 12-d-old or 14-d-old Super:C-repeat-binding factors (CBFs)-Myc plants for CBF protein immunoblots to study the effect of R18 on CBF3 degradation in planta.

Biochem/physiol Actions

C-repeat-binding factors (CBFs) protein -Myc transgenic plants, R18 is capable of reducing the degradation of CBF1 and CBF3.
R18 (PHCVPRDLSWLDLEANMCLP), a competitive inhibitor of 14-3-3 scaffolding proteins, may be used to study the roles and functions of 14-3-3 scaffolding proteins in cell signaling pathways involved in processes such as apoptosis, division, migration, stress response, and malignant transformation.
R18 is a competitive inhibitor of the 14-3-3 scaffolding proteins. It binds to the general binding groove of 14-3-3. R18 blocks 14-3-3 interaction with most or all of its client proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol Mackintosh
The Biochemical journal, 381(Pt 2), 329-342 (2004-05-29)
14-3-3 proteins exert an extraordinarily widespread influence on cellular processes in all eukaryotes. They operate by binding to specific phosphorylated sites on diverse target proteins, thereby forcing conformational changes or influencing interactions between their targets and other molecules. In these
14-3-3zeta binds a phosphorylated Raf peptide and an unphosphorylated peptide via its conserved amphipathic groove
Petosa C, et al.
The Journal of biological chemistry, 273(26), 16305-16310 (1998)
14-3-3 Dimers Probe the Assembly Status of Multimeric Membrane Proteins
Yuan H, et al.
Current Biology, 13(8), 638-646 (2003)
Plasma membrane CRPK1-mediated phosphorylation of 14-3-3 proteins induces their nuclear import to fine-tune CBF signaling during cold response
Liu Z, et al.
Molecular Cell, 66(1), 117-128 (2017)
Yuping Chen et al.
Science advances, 6(1), eaax5819-eaax5819 (2020-01-09)
Autophagy is an evolutionarily conserved catabolic process, which plays a vital role in removing misfolded proteins and clearing damaged organelles to maintain internal environment homeostasis. Here, we uncovered the checkpoint kinase 2 (CHK2)-FOXK (FOXK1 and FOXK2) axis playing an important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service