Skip to Content
Merck
All Photos(1)

Key Documents

SML0033

Sigma-Aldrich

Itopride hydrochloride

≥98% (HPLC)

Synonym(s):

N-[[4-[2-(Dimethylamino)ethoxy]phenyl]methyl]-3,4-dimethoxy benzamidehydrochloride, N-[p-[2-(dimethylamino)ethoxy]benzyl]veratramide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2O4 ·HCl
CAS Number:
Molecular Weight:
394.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥48 mg/mL

originator

Abbott

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(cc1OC)C(=O)NCc2ccc(OCCN(C)C)cc2

InChI

1S/C20H26N2O4.ClH/c1-22(2)11-12-26-17-8-5-15(6-9-17)14-21-20(23)16-7-10-18(24-3)19(13-16)25-4;/h5-10,13H,11-12,14H2,1-4H3,(H,21,23);1H

InChI key

ZTOUXLLIPWWHSR-UHFFFAOYSA-N

General description

Itopride hydrochloride is a new prokinetic drug.[1]

Biochem/physiol Actions

Antagonist of dopamine D2-receptor with anticholinesterase activity.
Itopride hydrochloride enhances the gastrointestinal motility by blocking the activity of dopamine on the D2 receptors, on the post-synaptic cholinergic nerves and by inducing the liberation of acetylcholine in the myenteric plexus. It also inhibits the hydrolysis of the released acetylcholine with the help of acetylcholinesterase.[1]
Itopride is an acetylcholinesterase (AChE) inhibitor and D2 dopamine receptor antagonist. Itopride is used as a gastroprokinetic agent.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Effect of HSR-803 on gastrointestinal motility].
H Iwanaga et al.
Nihon Heikatsukin Gakkai zasshi, 25(6), 313-315 (1989-12-01)
Sanjay Bansal et al.
Drug delivery, 23(2), 437-451 (2014-05-29)
The objectives of present studies were to develop the systematically optimized multiple-unit gastroretentive microballoons, i.e. hollow microspheres of itopride hydrochloride (ITH) employing quality by design (QbD)-based approach. Initially, the patient-centric QTPP and CQAs were earmarked, and preliminary studies were conducted
E Takahara et al.
Journal of chromatography, 576(1), 174-178 (1992-04-15)
A method based on high-performance liquid chromatography using column-switching is described for the simultaneous determination of HSR-803 and its metabolites in human serum and urine. The system uses a six-port valve with a Nucleosil CN pre-column for on-line sample clean-up
Comparative evaluation of the efficacy and tolerability of itopride hydrochloride and domperidone in patients with non-ulcer dyspepsia.
Sawant P, et al.
The Journal of the Association of Physicians of India, 52, 626-628 (2004)
Y Iwanaga et al.
Japanese journal of pharmacology, 56(3), 261-269 (1991-07-01)
To estimate the effect of a new gastroprokinetic agent, N-[4-[2-(dimethylamino)ethoxy]benzyl]-3,4-dimethoxybenzamide hydrochloride (HSR-803), on non-ulcer dyspepsia, the influence of HSR-803 on gastrointestinal propulsion was assayed in dogs, rats and mice in comparison with some gastroprokinetic agents. HSR-803 (30 mg/kg, p.o.) significantly

Questions

  1. Is the Itopride HCL CAS 122892-31-3 a primary standard grade or a reagent grade?

    1 answer
    1. This product is a research grade Itopride and not considered a primary or secondary standard. Currently, there is no primary standard product available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service