Skip to Content
Merck
All Photos(1)

Key Documents

SMB01000

Sigma-Aldrich

Grandiflorenic acid

≥90% (LC/MS-ELSD)

Synonym(s):

(4S,5R,9R)-5,9-Dimethyl-14-methylidenetetracyclo[11.2.1.01,10.04,9]hexadec-10-ene-5-carboxylic acid, Kaura-9(11),16-dien-18-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

300.44

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC(=O)[C@]1([C@@H]2[C@@](CCC1)(C3=CCC4CC3(CC2)CC4=C)C)C

InChI

1S/C20H28O2/c1-13-11-20-10-7-15-18(2,16(20)6-5-14(13)12-20)8-4-9-19(15,3)17(21)22/h6,14-15H,1,4-5,7-12H2,2-3H3,(H,21,22)/t14?,15-,18+,19+,20?/m0/s1

InChI key

RJIPNPHMQGDUBW-MZWQVCBVSA-N

General description

Grandiflorenic acid, a diterpenoid, is a natural product commonly derived from Wedelia trilobata, Sphagneticola trilobata and Siegesbeckia orientalis L. It exhibits various biological activities such as anti-inflammatory, anti-depressant, anti-fertility, and anti-microbial properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Grandiflorenic acid exhibits antibacterial, anti-inflammatory, antileishmanial and wound-healing properties.
Grandiflorenic acid shows moderate cytotoxic (IC50 15μM in RAW264.7 cells) and antiplasmodial activities (IC50 23μM against P. falciparum).
Grandiflorenic acid (GFA) shows promise as an anticancer agent by inhibiting cell proliferation and reducing cell size in breast (MCF7), lung (A549), and liver (HuH7.5) tumor cell lines. Further, it also shows effects on uterine motility throughout the estrus cycle suggesting it may have antifertility properties, possibly by affecting hormones and adrenergic pathways in the uterus.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amira Arciniegas et al.
Chemistry & biodiversity, 15(3), e1700529-e1700529 (2018-02-11)
The chemical study of Ageratina deltoidea afforded grandiflorenic acid (1), ent-kaurenoic acid (2), and eight benzylbenzoates (3 - 10), two of them, 3,5-dimethoxybenzyl 2,3,6-trimethoxybenzoate (5) and 4-(β-d-glucopyranosyloxy)-3-hydroxybenzyl 2,6-dimethoxybenzoate (9), described for the first time. In addition, the new sesquiterpene lactone deltoidin C
The in vitro effect of grandiflorenic acid and zoapatle aqueous crude extract upon spontaneous contractility of the rat uterus during oestrus cycle
Bejar E. et al.
Journal of Ethnopharmacology, 11, 87-97 (1984)
Bruna Taciane da Silva Bortoleti et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 46, 11-20 (2018-08-12)
American tegumentary leishmaniasis (ATL) is a zoonotic disease caused by protozoa of the genus Leishmania. The high toxicity, high costs and resistance of some strains to current drugs has prompted the search for therapeutic alternatives for the management of this
Grandiflorenic acid from Wedelia trilobata plant induces apoptosis and autophagy cell death in breast adenocarcinoma (MCF7), lung carcinoma (A549), and hepatocellular carcinoma (HuH7.5) cells lines
Fagundes TR, et al.
Toxicon, 217, 112-120 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service