Skip to Content
Merck
All Photos(1)

Key Documents

SMB00586

Sigma-Aldrich

Nimbolide

from Azadirachta indica, ≥98%

Synonym(s):

NSC-309909

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O7
CAS Number:
Molecular Weight:
466.52
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98%

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[H][C@@]12C[C@@H](C3=COC=C3)C(C)=C1[C@]([C@H](CC(OC)=O)[C@](C(C=C[C@]45C)=O)(C)[C@@]4([H])[C@@]6([H])OC5=O)(C)[C@]6([H])O2

InChI

1S/C27H30O7/c1-13-15(14-7-9-32-12-14)10-16-20(13)27(4)17(11-19(29)31-5)26(3)18(28)6-8-25(2)22(26)21(23(27)33-16)34-24(25)30/h6-9,12,15-17,21-23H,10-11H2,1-5H3/t15-,16-,17-,21-,22+,23-,25-,26+,27-/m1/s1

InChI key

JZIQWNPPBKFOPT-LSYMHUITSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nimbolide is a limonoid tetranortriterpenoid with an α,β-unsaturated ketone system, and a δ-lactone ring. It is usually isolated from Azadirachta indica (Neem).

Application

Nimbolide has been used as an anticancer agent to study its effects on the proliferation, migration, and invasion of human bladder cancer cell lines (EJ and 5637). It has also been used as a treatment to target RNF114 and trap PARP1 inhibitor(PARPi) and PARylation-dependent DNA repair factors.

Biochem/physiol Actions

Nimbolide exhibits anti-malarial, anti-bacterial, and anti-cancer activities. It has been shown to induce apoptosis in osteosarcoma cells and inhibits cell migration of these cells by modulating the expression of integrin αvβ5. In human renal carcinoma cells, nimbolide induced cell cycle arrest, DNA damage, and apoptosis.It has been shown to modulate autophagy and apoptosis in pancreatic cancer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Seung-Shick Shin et al.
Evidence-based complementary and alternative medicine : eCAM, 2019, 3753587-3753587 (2019-08-09)
Nimbolide, an active chemical constituent of Azadirachta indica, reportedly has several physiological effects. Here, we assessed novel anticancer effects of nimbolide against bladder cancer EJ and 5637 cells. Nimbolide treatment inhibited the proliferation of both bladder cancer cell lines with
Panawan Suttiarporn et al.
Molecules (Basel, Switzerland), 25(12) (2020-07-01)
Nimbolide, a limonoid present in leaves of the neem tree (Azadirachta indica), is an anticancer compound against a panel of human cancer cell lines. The rapid process of extraction and purification of the nimbolide from the leaves of neem tree
Nimbolide inhibits pancreatic cancer growth and metastasis through ROS-mediated apoptosis and inhibition of epithelial-to-mesenchymal transition.
Subramani R, et al.
Scientific Reports, 6, 19819-19819 (2016)
Yi-Hsien Hsieh et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(10), 7539-7547 (2015-04-29)
Nimbolide is a tetranortriterpenoid isolated from the leaves and flowers of Azadirachta indica which has been shown to exhibit anticancer, antioxidant, anti-inflammatory, and anti-invasive properties in a variety of cancer cells. However, the anti-tumor effect on human renal cell carcinoma
Chemopreventive and therapeutic effects of nimbolide in cancer: the underlying mechanisms.
Bodduluru L N, et al.
Toxicology in vitro, 28(5), 1026-1035 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service