Skip to Content
Merck
All Photos(1)

Key Documents

SMB00131

Sigma-Aldrich

3,5-Di-caffeoylquinic acid

≥95% (LC/MS-ELSD)

Synonym(s):

3,5-DCQA, 3,5-Dicaffeoylquinate, 3,5-Dicaffeoylquinic acid, 3,5-O-Dicaffeoylquinic acid, Isochlorogenic Acid A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](C[C@@](O)(C[C@H]1OC(=O)\C=C\c2ccc(O)c(O)c2)C(O)=O)OC(=O)\C=C\c3ccc(O)c(O)c3

InChI

1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(30)36-19-11-25(35,24(33)34)12-20(23(19)32)37-22(31)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-29,32,35H,11-12H2,(H,33,34)/b7-3+,8-4+/t19-,20-,23-,25+/m1/s1

InChI key

KRZBCHWVBQOTNZ-PSEXTPKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Dicaffeoylquinic acid is a phenolic acid found in the flower buds of Lonicera japonica Thunb.[1] It has been used in pharmacologic, as well as food, and cosmetic research.

Application

3,5-Di-caffeoylquinic acid has been used as a reference standard for quantification of the phenolic compound of Artemisia species using high-performance liquid chromatography with diode array detection (HPLC-DAD).[2][3] It has also been used as a reference standard to quantify the polyphenolic compounds found in Helianthus tuberosus L. (Jerusalem artichoke) using high-performance liquid chromatography-ultraviolet (HPLC-UV) technique.[4]

Biochem/physiol Actions

3,5-Di-caffeoylquinic acid exerts neuroprotective effects against in vitro retinal damage mediated by its antioxidant properties.[5] It is a potent maltase inhibitor.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

alpha-Glucosidase inhibitory activity by the flower buds of Lonicera japonica Thunb
Zhang Z, et al.,
Journal of functional foods, 5(3), 1253-1259 (2013)
Jiahui Chang et al.
Journal of ethnopharmacology, 194, 1032-1042 (2016-11-12)
Houshiheisan (HSHS), a classic traditional medicine prescription, has notable effects on patients with stroke AIM OF THE STUDY: To investigate the neurorestorative effects of HSHS on ischemic stroke and explore its mode of action. Focal cerebral ischemia models were induced
Sebastian E W Opitz et al.
Phytochemical analysis : PCA, 28(2), 106-114 (2016-12-23)
Coffee is a widely consumed beverage containing antioxidant active compounds. During roasting the phytochemical composition of the coffee bean changes dramatically and highly polymeric substances are produced. Besides chlorogenic acids that are already present in green coffee beans, melanoidins show
John Tung Chien et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 49, 1-10 (2018-09-16)
Non-small-cell lung cancer (NSCLC) is known to exhibit resistance to various therapeutic agents and become progressively incurable. Taraxacum formosanum is a medicinal Chinese herb that has been clinically used in Taiwan. However, the investigations of the effects of whole plant
Water extract of propolis and its main constituents, caffeoylquinic acid derivatives, exert neuroprotective effects via antioxidant actions
Nakajima Y, et al.,
Life Sciences, 80(4), 370-377 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service