Skip to Content
Merck
All Photos(1)

Key Documents

SCP0225

Sigma-Aldrich

Proteasome Substrate

≥95% (HPLC), lyophilized

Synonym(s):

Carbobenzoxy-Gly-Gly-Leu-7-amido-4-methylcoumarin, benzyloxycarbonyl-glycyl-glycyl-leucyl-7-amido-4-methylcoumarin, Z-GGL-AMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N4O7
Molecular Weight:
536.58
UNSPSC Code:
12352204
NACRES:
NA.32

product name

Proteasome Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥87%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Z-Gly-Gly-Leu-AMC

Application

Z-Gly-Gly-Leu-7-amido-4-methylcoumarin (Z-Gly-Gly-Leu-AMC) has been used as a substrate for proteasome peptidase to measure proteosome activities using spectrophotometer.

Biochem/physiol Actions

Z-Gly-Gly-Leu-7-amido-4-methylcoumarin (Z-Gly-Gly-Leu-AMC) is a fluorogenic peptide that is used in analysis of protease and peptidase activity of proteasomes. Z-GGL-AMC has been noted as a particular substrate for chymotrypsin-like activity. It has low solubility and precipitates at 100μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

István Nagy et al.
Journal of bacteriology, 185(2), 496-503 (2003-01-04)
In a proteasome-lacking mutant of Streptomyces coelicolor A3(2), an intracellular enzyme with chymotrypsin-like activity, absent from the wild type, was detected. Complementation that restored proteasome function did not suppress expression of the endopeptidase. Since the enzyme was not found in
M Rohrwild et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(12), 5808-5813 (1996-06-11)
We have isolated a new type of ATP-dependent protease from Escherichia coli. It is the product of the heat-shock locus hslVU that encodes two proteins: HslV, a 19-kDa protein similar to proteasome beta subunits, and HslU, a 50-kDa protein related
C P Ma et al.
The Journal of biological chemistry, 267(15), 10515-10523 (1992-05-25)
A protein that greatly stimulates the multiple peptidase activities of the 20 S proteasome (also known as macropain, the multicatalytic protease complex, and 20 S protease) has been purified from bovine red blood cells and from bovine heart. The activator
Raymond J. Deshaies
Ubiquitin and Protein Degradation, Part 1, 1 null
S Rathore et al.
Cell death & disease, 2, e231-e231 (2011-11-25)
The ATP-dependent ClpQY protease system in Plasmodium falciparum is a prokaryotic machinery in the parasite. In the present study, we have identified the complete ClpQY system in P. falciparum and elucidated its functional importance in survival and growth of asexual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service