SAB5702276
Anti-FITC/5-FAM/6-FAM Antibody, clone 6V10D5, Rabbit Monoclonal
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
biological source
rabbit
Quality Level
material
colorless
clone
6V10D5, monoclonal
form
liquid
concentration
1.00 mg/mL
color
colorless
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Immunogen
FITC
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Application
ELISA, WB
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service