Skip to Content
Merck
All Photos(10)

Key Documents

SAB4504449

Sigma-Aldrich

Anti-phospho-SAPK/JNK (pThr183) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JNK, Anti-JNK-46, Anti-JNK1, Anti-JNK1A2, Anti-JNK21B1/2, Anti-PRKM8, Anti-SAPK1, Anti-SAPK1c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:4000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr183)

Gene Information

Related Categories

Immunogen

The antiserum was produced against synthesized peptide derived from human SAPK/JNK around the phosphorylation site of Thr183.

Immunogen Range: 151-200

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunhua Wang et al.
Molecular medicine reports, 16(1), 922-928 (2017-06-01)
In the present study, the protective effect and the potential underlying mechanism of rutaecarpine (Ru) on renal ischemia reperfusion injury (IRI) in rats were investigated. A renal ischemia reperfusion mouse model was established. Ru at 30, 60 mg/kg administered intraperitoneally prior
Xiuli Zhang et al.
International journal of molecular medicine, 35(6), 1747-1754 (2015-04-16)
Zinc (Zn) as an essential dietary element has been indicated in a number of protein functions in the prevention of numerous types of epithelial-to-mesenchymal transition (EMT)-driven fibrosis in vivo. However, relatively little is known regarding its effect in the EMT
Xiuli Zhang et al.
Kidney & blood pressure research, 43(2), 500-512 (2018-04-09)
Evidence from our and other groups has demonstrated that zinc transporter 7 in SLC30 family (ZnT7) inhibited epithelial-to-mesenchymal transition (EMT) and apoptosis in rat peritoneal mesothelial cells (RPMCs) under high glucose (HG) concentration. In the present study, we investigated the
Jiao Zheng et al.
Cancer management and research, 11, 5231-5242 (2019-06-27)
Background: Hepatocellular carcinoma (HCC) is a particularly prevalent type of liver cancer and is one of the deadliest malignancies in Asia. Tangeretin is a biological compound extracted from traditional Chinese herbs and has been shown to have potential antitumour properties;
Yanfang Li et al.
Cardiovascular engineering and technology, 11(2), 219-227 (2020-01-10)
Recent studies have demonstrated that miRNAs play a vital role in regulating myocardial ischemia/reperfusion injury (MIRI). MiR-217 has been proven to be implicated in cardiac diseases such as chronic heart failure and cardiac myxoma. However, the role of miR-217 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service