Skip to Content
Merck
All Photos(4)

Key Documents

SAB4504108

Sigma-Aldrich

Anti-phospho-TIP60 (pSer86) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KAT5, Lysine acetyltransferase 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer86)

Gene Information

human ... KAT5(10524)

General description

The TIP60 (HIV1 (human immunodeficiency virus 1)-TAT interactive protein) gene is mapped to human chromosome 11q13.1. It is ubiquitously expressed and evolutionarily conserved. The encoded protein, Tip60 belongs to the MYST (Moz, Ybf2/Sas3, Sas2 and TIP60) family.

Immunogen

The antiserum was produced against synthesized peptide derived from human TIP60 around the phosphorylation site of Ser86.

Immunogen Range: 52-101

Application

TIP60 (HIV1-TATinteractive protein) possess acetyltransferase activity and participates in chromatin remodeling and DNA damage repair. TIP60 is involved in protein stabilization and protein function modulation. The presence of chromodomain and MYST domain in its structure enables acetylation of both histone and non-histone proteins. TIP60 in association with androgenic receptors and transcription factors can control gene transcription and cell cycle process. TIP60 mediates p53 action to induce apoptosis. TIP60 inhibits Hepatitis B virus infection. TIP60 acts as a haploinsufficient tumor suppressor.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TIP60 represses telomerase expression by inhibiting Sp1 binding to the TERT promoter
Rajagopalan D, et al.
PLoS Pathogens, 13(10), e1006681-e1006681 (2017)
Cellular functions of TIP60
Sapountzi V, et al.
The International Journal of Biochemistry & Cell Biology, 38(9), 1496-1509 (2006)
Interaction of the epigenetic integrator UHRF1 with the MYST domain of TIP60 inside the cell
Ashraf W, et al.
Journal of Experimental & Clinical Cancer Research, 36(1), 188-188 (2017)
TIP60 complex inhibits HBV transcription
Nishitsuji H, et al.
Journal of Virology, JVI-01788 (2018)
Marcella M Authiat et al.
Neural plasticity, 2022, 3172861-3172861 (2022-03-04)
Recently, we showed that DNA double-strand breaks (DSBs) are increased by the Aβ 42-amyloid peptide and decreased by all-trans retinoic acid (RA) in SH-SY5Y cells and C57BL/6J mice. The present work was aimed at investigating DSBs in cells and murine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service