Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503898

Sigma-Aldrich

Anti-phospho-Aurora Kinase (pThr288) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 45 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr288)

Gene Information

human ... AURKA(6790)

General description

Aurora kinase A (AURKA) belongs to the serine/threonine kinase family. It is localized to centrosomes. AURKA gene is located on human chromosome 20q13.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human Aurora Kinase around the phosphorylation site of Thr288.

Immunogen Range: 256-305

Application

Anti-phospho-Aurora Kinase (pThr288) antibody produced in rabbit has been used in immunofluorescence staining.

Biochem/physiol Actions

Aurora kinase A (AURKA) is involved in maintaining the stability of mitosis and chromosomes. During mitosis, AURKA participates in centrosome maturation control, mitotic entry, bipolar spindle formation, and segregation. AURKA also participates in chromosome alignment and cytokinesis. It controls several oncogenic signaling pathways. AURKA plays a major role in the self-renewal, reprogramming, and differentiation of stem cells. Knockdown of AURKA gene affects the maturation of the centrosome. Abnormality in the AURKA gene leads to aberrant cell division and aneuploidy, which in turn affects the stability of the genome and contributes to carcinogenesis. Overexpression of the AURKA gene is seen in gastrointestinal, pancreatic, and liver cancers. It can act as a candidate gene to predict colorectal adenocarcinoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Dicioccio et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 13(10), 1589-1594 (2004-10-07)
STK15 is a putative oncogene that codes for a centrosome-associated, serine/threonine kinase, the normal function of which is to ensure accurate segregation of chromosomes during mitosis. Amplification of STK15 has been reported in ovarian tumors, suggesting a role in ovarian
Ahmed Katsha et al.
Molecular cancer, 14, 106-106 (2015-05-20)
Gastrointestinal (GI) cancers are a major cause of cancer-related deaths. During the last two decades, several studies have shown amplification and overexpression of Aurora kinase A (AURKA) in several GI malignancies. These studies demonstrated that AURKA not only plays a
Hyun Min Koh et al.
Journal of pathology and translational medicine, 51(1), 32-39 (2016-12-26)
Aurora kinase A (AURKA), or STK15/BTAK, is a member of the serine/threonine kinase family and plays important roles in mitosis and chromosome stability. This study investigated the clinical significance of AURKA expression in colorectal cancer patients in Korea. AURKA protein
Yan-Ru Li et al.
Aging, 11(4), 1110-1128 (2019-02-21)
Tight control of energy metabolism is essential for normal cell function and organism survival. PKM (pyruvate kinase, muscle) isoforms 1 and 2 originate from alternative splicing of PKM pre-mRNA. They are key enzymes in oxidative phosphorylation and aerobic glycolysis, respectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service