Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503544

Sigma-Aldrich

Anti-Claudin 2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CLD2, CLDN2, SP82, claudin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 24 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN2(9075)

General description

Anti-Claudin 2 Antibody detects endogenous levels of total Claudin 2 protein.
Claudin-2 is one of the pore-forming claudins. It is a member of the claudin family of proteins. Claudin-2 is present in the proximal tubules. It is mapped to human chromosome Xq22.

Immunogen

The antiserum was produced against synthesized peptide derived from human Claudin 2.

Immunogen Range: 181-230

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

The tetraspan membrane protein encoded by claudin multigene controls paracellular permeability and maintaining cell polarity in epithelial and endothelial cell sheets. In MDCK (madin-darby canine kidney cells) II cells, claudin-2 participates in the generation of aqueous pores with high conductance.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inga B Christensen et al.
Frontiers in physiology, 4, 344-344 (2013-12-19)
The choroid plexus epithelium (CPE) has served as a model-epithelium for cell polarization and transport studies and plays a crucial role for cerebrospinal fluid (CSF) production. The normal luminal membrane expression of Na(+),K(+)-ATPase, aquaporin-1 and Na(+)/H(+) exchanger 1 in the
Tumor necrosis factor-? induces a biphasic change in claudin-2 expression in tubular epithelial cells: role in barrier functions
Amoozadeh Y, et al.
American Journal of Physiology. Cell Physiology, 309(1), C38-C50 (2015)
Inhibition of Autophagic Degradation Process Contributes to Claudin-2 Expression Increase and Epithelial Tight Junction Dysfunction in TNF-? Treated Cell Monolayers
Zhang C, et al.
International Journal of Molecular Sciences, 18(1) (2017)
Conversion of Zonulae Occludentes from Tight to Leaky Strand Type by Introducing Claudin-2 into Madin-Darby Canine Kidney I Cells
Furuse M, et al.
The Journal of Cell Biology, 153(2), 263-272 (2001)
The claudins
Lal-Nag M and Morin P J
Genome Biology (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service