Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503479

Sigma-Aldrich

Anti-FGF18 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FGF-18, Fibroblast growth factor 18, Zfgf5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF18(8817)

General description

Anti-FGF18 Antibody detects endogenous levels of total FGF18 protein.
FGF18 (fibroblast growth factor 18) is a member of the paracrine-acting FGF8 subfamily. It is expressed in the perichondrium and joints of developing long bones. FGF18 is a 21.2 kDa glycosylated secretory protein, that is structurally similar to FGF8 and FGF17. It is mapped to human chromosome 5q35.1. Anti-FGF18 Antibody detects endogenous levels of total FGF18 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human FGF18.

Immunogen Range: 151-200

Biochem/physiol Actions

FGF18 (fibroblast growth factor 18) plays an important role in the skeletal development of the embryonic and adult organism. It is associated with colorectal and ovarian cancer. FGF18 participates in cell proliferation and differentiation during osteogenesis and chondrogenesis. It is essential for the growth of bone, cartilage, hair, cardiovasculature and alveolus.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of fibroblast growth factor 18 (FGF18).
Alan B, et al.
Protein & cell, 5(5), 343?347-343?347 (2014)
FGF18 is required for normal cell proliferation and differentiation during osteogenesis and chondrogenesis.
Ohbayashi N, et al.
Genes & Development, 16(7), 870-879 (2002)
Nobuyuki Itoh et al.
Trends in genetics : TIG, 20(11), 563-569 (2004-10-12)
Fibroblast growth factors (Fgfs) and Fgf receptors (Fgfrs) comprise a signaling system that is conserved throughout metazoan evolution. Twenty-two Fgfs and four Fgfrs have been identified in humans and mice. During evolution, the Fgf family appears to have expanded in
Wei Wei et al.
The Journal of clinical investigation, 123(10), 4435-4448 (2013-09-11)
High-throughput genomic technologies have identified biomarkers and potential therapeutic targets for ovarian cancer. Comprehensive functional validation studies of the biological and clinical implications of these biomarkers are needed to advance them toward clinical use. Amplification of chromosomal region 5q31-5q35.3 has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service